siRNA Details
Gene Symbol: NEU4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167599.2 | NP_001161071.1 | 2 | 569 | ||
NM_001167600.2 | NP_001161072.1 | 2 | 246 | |||
NM_001167601.2 | NP_001161073.1 | 2 | 249 | |||
NM_001167602.2 | NP_001161074.1 | 3 | 747 | |||
NM_080741.3 | NP_542779.2 | 2 | 566 | |||
GenBank mRNA | AJ277883.1 | - | 1 | 33 | ||
AK025617.1 | - | 2 | 118 | |||
AK091038.1 | - | 3 | 744 | |||
AK096992.1 | - | 2 | 121 | |||
AK289421.1 | - | 2 | 123 | |||
AK290076.1 | - | 2 | 106 | |||
AK298403.1 | - | 2 | 118 | |||
AK300969.1 | - | 2 | 119 | |||
AY203952.1 | - | 2 | 102 | |||
BC012899.1 | - | 2 | 520 | 4156395 | ||
BC105974.1 | - | 2 | 82 | 4944243 | ||
BC117214.1 | - | 2 | 210 | 40125765 | ||
BQ651855.1 | - | 2 | 249 | |||
DB173219.1 | - | 2 | 503 | |||
DC412650.1 | - | 2 | 486 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
