siRNA Details
Gene Symbol: TRMT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020810.3 | NP_065861.3 | 5 | 4318 | ||
XM_005267916.3 | XP_005267973.2 | 5 | 4348 | |||
GenBank mRNA | AB037814.1 | - | 4 | 4190 | ||
BC063551.1 | - | 1 | 373 | |||
BC136606.1 | - | 5 | 4233 | 9020596 | ||
BC136607.1 | - | 5 | 4233 | 9020597 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
