siRNA Details
Gene Symbol: PDDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318818.1 | NP_001305747.1 | 7 | 2464 | ||
NM_001318820.1 | NP_001305749.1 | 6 | 2522 | |||
NM_001318821.1 | NP_001305750.1 | 7 | 2630 | |||
NM_001318822.1 | NP_001305751.1 | 5 | 2445 | |||
NM_001318823.1 | NP_001305752.1 | 6 | 2553 | |||
NM_001318824.1 | NP_001305753.1 | 7 | 2433 | |||
NM_182612.3 | NP_872418.1 | 8 | 2541 | |||
NR_134867.1 | 7 | 2572 | ||||
NR_134868.1 | 8 | 2635 |
Gene Symbol: ABCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017018993.1 | XP_016874482.1 | 7 | 4552 |
Gene Symbol: ACTR3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209174.1 | - | 1 | 2844 |
Gene Symbol: AHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001621.4 | NP_001612.1 | 11 | 4785 | ||
GenBank mRNA | AA329775.1 | - | 1 | 212 | ||
L19872.1 | - | 11 | 4554 |
Gene Symbol: ALKBH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX537699.1 | - | 4 | 2570 |
Gene Symbol: ALPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017008644.1 | XP_016864133.1 | 1 | 1069 |
Gene Symbol: BMS1P20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX641141.1 | - | 5 | 1661 |
Gene Symbol: C14orf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK309499.1 | - | 3 | 891 | ||
AK310120.1 | - | 4 | 1038 |
Gene Symbol: C16orf86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255952.4 | XP_005256009.1 | 4 | 2992 |
Gene Symbol: CBX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127321.1 | NP_001120793.1 | 5 | 4303 | ||
NM_001127322.1 | NP_001120794.1 | 5 | 4501 | |||
NM_012117.2 | NP_036249.1 | 5 | 4349 | |||
GenBank mRNA | AK023520.1 | - | 1 | 476 | ||
AK095108.1 | - | 1 | 2761 |
Gene Symbol: DCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000555.3 | NP_000546.2 | 7 | 6459 | ||
NM_001195553.1 | NP_001182482.1 | 7 | 6179 | |||
NM_178151.2 | NP_835364.1 | 7 | 6288 | |||
NM_178152.2 | NP_835365.1 | 7 | 6176 | |||
NM_178153.2 | NP_835366.1 | 7 | 6161 | |||
XM_011530878.2 | XP_011529180.1 | 7 | 6136 | |||
XM_011530879.2 | XP_011529181.1 | 6 | 6053 | |||
XM_011530880.2 | XP_011529182.1 | 6 | 6038 | |||
XM_017029312.1 | XP_016884801.1 | 7 | 6244 | |||
GenBank mRNA | AJ003112.1 | - | 7 | 6469 | ||
AK299822.1 | - | 3 | 1252 |
Gene Symbol: DNAJB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017679.1 | XP_016873168.1 | 6 | 1684 |
Gene Symbol: EIF4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR749455.1 | - | 2 | 1936 |
Gene Symbol: FAM131A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024378.1 | - | 1 | 593 | ||
AK126656.1 | - | 2 | 1819 |
Gene Symbol: FKTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014476.1 | XP_016869965.1 | 7 | 768 | ||
XM_017014477.1 | XP_016869966.1 | 6 | 633 | |||
XM_017014478.1 | XP_016869967.1 | 8 | 835 | |||
XM_017014479.1 | XP_016869968.1 | 5 | 532 | |||
GenBank mRNA | BC117700.1 | - | 5 | 560 | 40073634 |
Gene Symbol: HTR5A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038945.1 | 2 | 2102 | |||
GenBank mRNA | BC031272.1 | - | 2 | 2070 | ||
BC045795.1 | - | 2 | 2073 |
Gene Symbol: IFT81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091549.1 | - | 18 | 1797 |
Gene Symbol: KRBOX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129898.1 | NP_001123370.1 | 6 | 1517 | ||
NM_001129899.1 | NP_001123371.1 | 7 | 1561 | |||
NM_001129900.1 | NP_001123372.1 | 7 | 1546 | |||
NM_017776.2 | NP_060246.2 | 6 | 1502 | |||
GenBank mRNA | AK000351.1 | - | 6 | 1369 | ||
AK096413.1 | - | 6 | 1411 | |||
AK097159.1 | - | 1 | 2387 |
Gene Symbol: LGMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008530.2 | NP_001008530.1 | 2 | 249 | ||
XM_017021466.1 | XP_016876955.1 | 1 | 24 | |||
XM_017021467.1 | XP_016876956.1 | 1 | 35 | |||
GenBank mRNA | BX161434.1 | - | 2 | 159 |
Gene Symbol: LINC00632 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DB307877.1 | - | 1 | 52 |
Gene Symbol: LINC01012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038292.1 | 3 | 1389 | |||
GenBank mRNA | BC035101.1 | - | 3 | 1390 |
Gene Symbol: LOC100130442 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131364.1 | - | 1 | 17 |
Gene Symbol: LOC101409256 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102424.1 | 1 | 1194 |
Gene Symbol: LOC101926955 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC052952.1 | - | 4 | 3583 |
Gene Symbol: LOC101930105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097803.1 | - | 4 | 1728 |
Gene Symbol: LOC102724392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134268.1 | 3 | 935 | |||
GenBank mRNA | AK123868.1 | - | 2 | 1105 | ||
AK310013.1 | - | 3 | 935 |
Gene Symbol: LOC102724580 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136300.1 | 1 | 150 |
Gene Symbol: LOC102725138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021789.1 | - | 1 | 22 |
Gene Symbol: LOC643623 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038906.1 | 4 | 1323 | |||
GenBank mRNA | BC035066.1 | - | 4 | 1330 |
Gene Symbol: MEAF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270875.1 | NP_001257804.1 | 7 | 3763 | ||
NM_001270876.1 | NP_001257805.1 | 6 | 3436 | |||
NM_022756.5 | NP_073593.2 | 8 | 3793 | |||
NR_073090.1 | 9 | 3851 | ||||
NR_073091.1 | 8 | 3821 | ||||
NR_073092.1 | 9 | 3882 |
Gene Symbol: MRS2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024072.2 | 1 | 39 | |||
GenBank mRNA | AY333753.1 | - | 40 |
Gene Symbol: MSH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097233.1 | - | 1 | 103 |
Gene Symbol: NCKAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092189.1 | - | 7 | 1317 |
Gene Symbol: NEK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193533.1 | NP_001180462.1 | 7 | 1339 | ||
NM_003157.4 | NP_003148.2 | 8 | 1606 | |||
XM_011534039.2 | XP_011532341.1 | 8 | 1603 | |||
XM_011534040.2 | XP_011532342.1 | 8 | 1603 | |||
XM_017007085.1 | XP_016862574.1 | 8 | 1603 | |||
GenBank mRNA | AK294165.1 | - | 7 | 1246 | ||
AK312420.1 | - | 8 | 1594 | |||
EF560744.1 | - | 8 | 1602 | |||
L20321.1 | - | 8 | 1581 |
Gene Symbol: OAZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648172.1 | - | 1 | 1860 |
Gene Symbol: OSBPL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF318344.1 | - | 1 | 2126 |
Gene Symbol: PCDHB16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB046841.1 | - | 1 | 4068 |
Gene Symbol: PHACTR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048183.1 | NP_001041648.1 | 14 | 5381 | ||
NM_023923.3 | NP_076412.3 | 13 | 5364 | |||
GenBank mRNA | AK024663.1 | - | 4 | 1033 | ||
BC021247.2 | - | 14 | 3250 | 4577873 |
Gene Symbol: PLPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124204.1 | - | 1 | 870 |
Gene Symbol: PROSER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124144.1 | - | 1 | 531 |
Gene Symbol: RAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK122742.1 | - | 1 | 581 |
Gene Symbol: RARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF118086.2 | - | 1 | 1051 |
Gene Symbol: RASGRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003759.1 | XP_016859248.1 | 3 | 924 | ||
XM_017003760.1 | XP_016859249.1 | 2 | 1396 | |||
GenBank mRNA | AK125551.1 | - | 3 | 971 |
Gene Symbol: RBFADN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX537710.1 | - | 1 | 535 |
Gene Symbol: RIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090451.1 | - | 3 | 3975 |
Gene Symbol: SDR42E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005256257.3 | XP_005256314.1 | 4 | 10670 | ||
XM_011523471.2 | XP_011521773.1 | 3 | 10479 | |||
XM_011523472.2 | XP_011521774.1 | 3 | 10468 |
Gene Symbol: SKIDA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207371.3 | NP_997254.3 | 4 | 5903 | ||
XM_005252450.3 | XP_005252507.1 | 5 | 5012 | |||
XM_011519480.2 | XP_011517782.1 | 5 | 5009 | |||
XM_017016225.1 | XP_016871714.1 | 5 | 5803 | |||
XM_017016226.1 | XP_016871715.1 | 5 | 5806 | |||
GenBank mRNA | AK127130.1 | - | 2 | 2934 | ||
AL834446.1 | - | 1 | 2616 |
Gene Symbol: SLC26A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057276.1 | - | 7 | 1135 |
Gene Symbol: SLC35G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006713773.3 | XP_006713836.1 | 1 | 1045 |
Gene Symbol: SLC36A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131483.1 | - | 1 | 346 |
Gene Symbol: SLFNL1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037868.1 | 4 | 4063 |
Gene Symbol: TRNAU1AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF118082.2 | - | 1 | 61 |
Gene Symbol: UIMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK304794.1 | - | 4 | 959 |
Gene Symbol: ULK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014683.3 | NP_055498.3 | 27 | 7033 | ||
GenBank mRNA | AL080134.1 | - | 1 | 990 |
Gene Symbol: USP9X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039590.2 | NP_001034679.2 | 45 | 10443 | ||
NM_001039591.2 | NP_001034680.2 | 45 | 10395 | |||
XM_005272675.4 | XP_005272732.1 | 45 | 10673 |
Gene Symbol: WDR5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019069.3 | NP_061942.2 | 1 | 2474 | ||
GenBank mRNA | BC043494.1 | - | 1 | 2063 | 6056052 |
Gene Symbol: ZKSCAN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242894.1 | NP_001229823.1 | 7 | 2641 | ||
NM_001242895.1 | NP_001229824.1 | 5 | 2060 | |||
NM_024493.3 | NP_077819.2 | 6 | 2524 | |||
XM_005249423.3 | XP_005249480.1 | 5 | 2690 | |||
XM_006715215.2 | XP_006715278.1 | 6 | 2422 | |||
XM_006715216.2 | XP_006715279.1 | 6 | 2437 | |||
XM_006715218.3 | XP_006715281.1 | 5 | 1988 | |||
XM_017011320.1 | XP_016866809.1 | 7 | 2518 | |||
GenBank mRNA | AK091225.1 | - | 5 | 2992 |
Gene Symbol: ZNF490 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020714.2 | NP_065765.1 | 5 | 5429 | ||
GenBank mRNA | AB033024.1 | - | 5 | 5421 | ||
BC038586.1 | - | 5 | 5410 | 5587423 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
