siRNA Details
Gene Symbol: POM121L8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024583.1 | 7 | 4394 | |||
GenBank mRNA | AK292222.1 | - | 4 | 1278 | ||
AL117485.1 | - | 5 | 2345 | |||
BC050331.1 | - | 2 | 578 | |||
BC068032.1 | - | 2 | 439 | |||
BC090873.1 | - | 2 | 299 |
Gene Symbol: LOC102725072 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135922.1 | 6 | 3333 | |||
GenBank mRNA | BC051721.1 | - | 6 | 3333 |
Gene Symbol: POM121L10P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024593.1 | 6 | 7532 | |||
GenBank mRNA | BX648116.1 | - | 5 | 2624 |
Gene Symbol: POM121L1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024591.1 | 10 | 3052 |
Gene Symbol: POM121L9P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003714.1 | 7 | 3280 | |||
GenBank mRNA | AL117401.1 | - | 7 | 3074 | ||
AL117484.1 | - | 4 | 1476 | |||
BC063879.1 | - | 2 | 777 | |||
BC086868.1 | - | 3 | 1602 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
