siRNA Details
Gene Symbol: OR2L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004687.1 | NP_001004687.1 | 1 | 324 | ||
GenBank mRNA | BC137039.1 | - | 1 | 344 | 9021035 | |
BC137040.1 | - | 1 | 344 | 9021036 |
Gene Symbol: OR2L8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001963.1 | NP_001001963.1 | 1 | 324 | ||
GenBank mRNA | BC140751.1 | - | 1 | 405 | 9021619 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
