siRNA Details
Gene Symbol: BMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014753.3 | NP_055568.3 | 17 | 3146 | ||
XM_005271846.3 | XP_005271903.1 | 17 | 3021 | |||
XM_005271848.3 | XP_005271905.1 | 16 | 2688 | |||
XM_011540402.2 | XP_011538704.1 | 17 | 2877 | |||
XM_011540403.2 | XP_011538705.1 | 9 | 1680 | |||
GenBank mRNA | BC043345.1 | - | 17 | 2841 | 6062924 | |
BC150252.1 | - | 16 | 2815 | 9007274 | ||
D80009.1 | - | 16 | 2815 |
Gene Symbol: BMS1P15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC107108.2 | - | 2 | 190 |
Gene Symbol: BMS1P20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027293.2 | 5 | 493 | |||
GenBank mRNA | AK090629.1 | - | 5 | 433 | ||
BC133659.1 | - | 2 | 190 | |||
BX641141.1 | - | 5 | 542 | |||
DB021899.1 | - | 5 | 494 |
Gene Symbol: LOC101929959 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011518094.2 | XP_011516396.1 | 4 | 784 | ||
XM_017015376.1 | XP_016870865.1 | 5 | 698 | |||
XM_017015377.1 | XP_016870866.1 | 5 | 698 | |||
XM_017015378.1 | XP_016870867.1 | 5 | 698 | |||
XM_017015379.1 | XP_016870868.1 | 5 | 698 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
