siRNA Details
Gene Symbol: ZNF320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207333.2 | NP_997216.2 | 4 | 2064 | ||
XM_006723059.3 | XP_006723122.1 | 5 | 2110 | |||
XM_017026393.1 | XP_016881882.1 | 6 | 2355 | |||
XM_017026394.1 | XP_016881883.1 | 6 | 2280 | |||
XM_017026395.1 | XP_016881884.1 | 6 | 2428 | |||
XM_017026396.1 | XP_016881885.1 | 5 | 2197 | |||
XM_017026397.1 | XP_016881886.1 | 5 | 2202 | |||
XM_017026398.1 | XP_016881887.1 | 5 | 2081 | |||
XM_017026399.1 | XP_016881888.1 | 4 | 2074 | |||
XM_017026400.1 | XP_016881889.1 | 4 | 1994 | |||
GenBank mRNA | AK096348.1 | - | 1 | 730 | ||
AL831902.1 | - | 1 | 1262 | |||
CR933622.1 | - | 2 | 1850 |
Gene Symbol: GTF2H3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CA306634.1 | - | 1 | 588 |
Gene Symbol: HTR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209129.1 | - | 1 | 4926 |
Gene Symbol: LOC101928215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK075019.1 | - | 2 | 445 |
Gene Symbol: NICN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032316.3 | NP_115692.1 | 6 | 1899 |
Gene Symbol: NUDCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX538290.1 | - | 4 | 7609 |
Gene Symbol: NUP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198887.2 | NP_942590.1 | 8 | 1878 | ||
XM_005266960.4 | XP_005267017.1 | 8 | 3108 | |||
XM_005266961.3 | XP_005267018.1 | 9 | 1928 | |||
XM_005266962.3 | XP_005267019.1 | 8 | 4148 | |||
XM_011535799.2 | XP_011534101.1 | 8 | 3190 | |||
GenBank mRNA | AK023349.1 | - | 3 | 1064 | ||
BC040121.1 | - | 6 | 1523 | |||
BC046637.1 | - | 1 | 1626 | |||
BC047539.1 | - | 1 | 2787 | |||
BC065028.1 | - | 8 | 1802 | 6042215 |
Gene Symbol: RBFADN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX537710.1 | - | 1 | 588 |
Gene Symbol: SENP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011512544.2 | XP_011510846.1 | 2 | 234 | ||
XM_017005870.1 | XP_016861359.1 | 2 | 189 |
Gene Symbol: SPIRE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC011119.1 | - | 3 | 1585 |
Gene Symbol: TEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB051442.1 | - | 1 | 3050 |
Gene Symbol: ZNF440 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152357.2 | NP_689570.2 | 4 | 3219 | ||
XM_005259731.3 | XP_005259788.1 | 4 | 3279 | |||
XM_017026253.1 | XP_016881742.1 | 3 | 3604 | |||
XM_017026254.1 | XP_016881743.1 | 3 | 3097 | |||
GenBank mRNA | AK092559.1 | - | 1 | 1315 | ||
AK095252.1 | - | 4 | 3220 |
Gene Symbol: ZNF468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008801.1 | NP_001008801.1 | 4 | 3288 | ||
NM_001277120.1 | NP_001264049.1 | 5 | 3406 | |||
NR_102299.1 | 6 | 3661 | ||||
NR_102300.1 | 6 | 3642 | ||||
NR_102301.1 | 5 | 3543 | ||||
XM_017027443.1 | XP_016882932.1 | 5 | 3503 | |||
XM_017027444.1 | XP_016882933.1 | 6 | 3621 | |||
GenBank mRNA | AK023558.1 | - | 1 | 1344 | ||
AK025501.1 | - | 1 | 1926 | |||
AY442330.1 | - | 4 | 3285 | |||
AY442331.1 | - | 5 | 3403 | |||
BC063143.1 | - | 1 | 1589 | |||
CR749381.1 | - | 1 | 2382 | |||
CR936599.1 | - | 4 | 3253 | |||
CR936695.1 | - | 4 | 3257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
