siRNA Details
Gene Symbol: REPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080975.1 | NP_001074444.1 | 18 | 4035 | ||
NM_004726.2 | NP_004717.2 | 18 | 4038 | |||
XM_005274625.3 | XP_005274682.1 | 16 | 3819 | |||
XM_005274626.3 | XP_005274683.1 | 16 | 3816 | |||
XM_017029958.1 | XP_016885447.1 | 16 | 3816 | |||
GenBank mRNA | AB208937.1 | - | 16 | 1898 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
