siRNA Details
Gene Symbol: NPIPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130464.2 | NP_569731.2 | 3 | 234 | ||
GenBank mRNA | AB209632.1 | - | 2 | 2801 | ||
AK295170.1 | - | 3 | 233 | |||
AK304162.1 | - | 1 | 16 | |||
BC036263.1 | - | 3 | 234 | 3903371 | ||
BC094882.1 | - | 2 | 161 | 3093162 | ||
DC342292.1 | - | 2 | 338 | |||
KF175175.1 | - | 2 | 101 | |||
KF175176.1 | - | 2 | 158 | |||
KF175177.1 | - | 2 | 158 | |||
KF175178.1 | - | 2 | 59 | |||
KF175179.1 | - | 2 | 101 | |||
KF175180.1 | - | 2 | 101 | |||
KF175181.1 | - | 2 | 245 | |||
KF175182.1 | - | 2 | 101 | |||
KF175185.1 | - | 2 | 101 | |||
KF175187.1 | - | 2 | 101 | |||
KF175188.1 | - | 2 | 62 | |||
KF175189.1 | - | 2 | 62 | |||
KF175190.1 | - | 2 | 101 | |||
KF175191.1 | - | 2 | 62 | |||
KF175192.1 | - | 2 | 62 | |||
KF175193.1 | - | 2 | 65 | |||
KF175194.1 | - | 2 | 65 | |||
KF175196.1 | - | 2 | 62 | |||
KF175197.1 | - | 2 | 62 | |||
KF175198.1 | - | 2 | 158 | |||
KF175199.1 | - | 2 | 158 | |||
KF175200.1 | - | 2 | 158 | |||
KF175201.1 | - | 2 | 101 | |||
KF175202.1 | - | 2 | 62 | |||
KF175203.1 | - | 2 | 158 | |||
KF175205.1 | - | 2 | 158 | |||
KF175206.1 | - | 2 | 158 | |||
KF175215.1 | - | 4 | 420 | |||
KF175216.1 | - | 3 | 435 | |||
KF175223.1 | - | 2 | 158 | |||
KF175224.1 | - | 2 | 101 |
Gene Symbol: LOC105376752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522797.2 | XP_011521099.1 | 2 | 232 |
Gene Symbol: LOC613037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321892.1 | NP_001308821.1 | 4 | 351 | ||
XM_017023514.1 | XP_016879003.1 | 4 | 640 | |||
XM_017023515.1 | XP_016879004.1 | 5 | 659 | |||
XM_017023516.1 | XP_016879005.1 | 4 | 686 | |||
XM_017023517.1 | XP_016879006.1 | 4 | 417 | |||
XM_017023518.1 | XP_016879007.1 | 4 | 505 | |||
XM_017023519.1 | XP_016879008.1 | 4 | 562 | |||
GenBank mRNA | BQ433543.1 | - | 274 | |||
DB231720.1 | - | 3 | 220 |
Gene Symbol: LOC729978 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG025764.1 | - | 3 | 251 |
Gene Symbol: NPIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006985.2 | NP_008916.2 | 2 | 101 | ||
XM_011522748.2 | XP_011521050.1 | 2 | 198 | |||
XM_011522749.2 | XP_011521051.1 | 3 | 310 | |||
XM_011522750.2 | XP_011521052.1 | 2 | 224 | |||
XM_011522751.1 | XP_011521053.1 | 2 | 197 | |||
XM_017023891.1 | XP_016879380.1 | 3 | 310 | |||
GenBank mRNA | AF132984.1 | - | 2 | 101 | ||
AK160377.1 | - | 21 | 4288 | |||
BE253379.1 | - | 3 | 369 | |||
KF175165.1 | - | 2 | 137 | |||
KF175167.1 | - | 2 | 137 | |||
KF175169.1 | - | 2 | 101 | |||
KF175208.1 | - | 2 | 248 | |||
KF175209.1 | - | 3 | 319 | |||
KF175225.1 | - | 2 | 101 |
Gene Symbol: NPIPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277324.1 | NP_001264253.1 | 2 | 101 | ||
XM_005255489.3 | XP_005255546.1 | 4 | 2520 | |||
XM_005255492.4 | XP_005255549.1 | 2 | 505 | |||
XM_006720917.3 | XP_006720980.2 | 4 | 2564 | |||
XM_011522587.1 | XP_011520889.1 | 4 | 2564 | |||
XM_011522593.1 | XP_011520895.1 | 4 | 2564 | |||
XM_011522594.2 | XP_011520896.1 | 4 | 2564 | |||
XM_011522595.1 | XP_011520897.1 | 4 | 2564 | |||
XM_011522596.1 | XP_011520898.1 | 2 | 128 | |||
XM_011522597.1 | XP_011520899.1 | 4 | 2564 |
Gene Symbol: NPIPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277323.1 | NP_001264252.1 | 2 | 101 | ||
XM_005255484.3 | XP_005255541.1 | 3 | 189 | |||
XM_005255485.4 | XP_005255542.1 | 2 | 505 | |||
XM_011522576.2 | XP_011520878.2 | 4 | 2554 | |||
XM_011522577.2 | XP_011520879.2 | 4 | 2475 | |||
XM_011522578.2 | XP_011520880.2 | 2 | 197 | |||
XM_011522580.2 | XP_011520882.2 | 2 | 128 | |||
XM_011522581.1 | XP_011520883.1 | 4 | 2564 | |||
XM_011522583.2 | XP_011520885.1 | 4 | 2564 | |||
XM_011522584.1 | XP_011520886.1 | 4 | 2564 | |||
XM_011522585.1 | XP_011520887.1 | 2 | 128 | |||
XM_011522586.1 | XP_011520888.1 | 4 | 2564 | |||
XM_017023553.1 | XP_016879042.1 | 4 | 2564 | |||
XM_017023554.1 | XP_016879043.1 | 3 | 2477 |
Gene Symbol: NPIPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277325.1 | NP_001264254.1 | 2 | 101 | ||
XM_011522329.2 | XP_011520631.1 | 4 | 512 | |||
XM_011522336.2 | XP_011520638.1 | 4 | 505 | |||
XM_011522337.1 | XP_011520639.1 | 2 | 128 | |||
GenBank mRNA | EL734608.1 | - | 2 | 101 | ||
KF175168.1 | - | 2 | 98 | |||
KF175170.1 | - | 2 | 101 | |||
KF175171.1 | - | 2 | 95 | |||
KF175172.1 | - | 2 | 101 |
Gene Symbol: NPIPA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282507.1 | NP_001269436.1 | 2 | 101 | ||
XM_005255735.4 | XP_005255792.1 | 2 | 223 | |||
XM_011522339.2 | XP_011520641.1 | 2 | 149 | |||
XM_017022833.1 | XP_016878322.1 | 3 | 174 | |||
XM_017022834.1 | XP_016878323.1 | 3 | 226 | |||
XM_017022835.1 | XP_016878324.1 | 3 | 287 | |||
XM_017022836.1 | XP_016878325.1 | 2 | 94 | |||
XM_017022837.1 | XP_016878326.1 | 3 | 174 | |||
XM_017022838.1 | XP_016878327.1 | 2 | 569 | |||
XM_017022839.1 | XP_016878328.1 | 3 | 174 | |||
XM_017022840.1 | XP_016878329.1 | 2 | 114 |
Gene Symbol: NPIPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282511.1 | NP_001269440.1 | 2 | 101 | ||
XM_005255738.4 | XP_005255795.1 | 2 | 223 | |||
XM_006720830.3 | XP_006720893.1 | 2 | 1121 | |||
XM_011522342.2 | XP_011520644.1 | 2 | 150 | |||
XM_017022841.1 | XP_016878330.1 | 3 | 174 | |||
XM_017022842.1 | XP_016878331.1 | 3 | 226 | |||
XM_017022843.1 | XP_016878332.1 | 3 | 287 | |||
XM_017022844.1 | XP_016878333.1 | 2 | 94 | |||
XM_017022845.1 | XP_016878334.1 | 3 | 174 | |||
XM_017022846.1 | XP_016878335.1 | 3 | 174 | |||
XM_017022847.1 | XP_016878336.1 | 2 | 114 | |||
GenBank mRNA | BQ101079.1 | - | 2 | 147 |
Gene Symbol: NPIPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310137.1 | NP_001297066.1 | 2 | 158 | ||
GenBank mRNA | BF592449.1 | - | 2 | 144 | ||
KF175195.1 | - | 2 | 62 | |||
KF175204.1 | - | 2 | 101 |
Gene Symbol: NPIPB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310148.1 | NP_001297077.1 | 3 | 383 | ||
XM_017023244.1 | XP_016878733.1 | 2 | 2394 | |||
XM_017023245.1 | XP_016878734.1 | 3 | 2231 | |||
XM_017023246.1 | XP_016878735.1 | 3 | 2152 | |||
XM_017023247.1 | XP_016878736.1 | 2 | 385 |
Gene Symbol: NPIPB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135865.1 | NP_001129337.1 | 2 | 437 | ||
GenBank mRNA | AK296338.1 | - | 2 | 278 | ||
DC309028.1 | - | 2 | 437 |
Gene Symbol: PKD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036447.1 | 27 | 5406 | |||
GenBank mRNA | AK124516.1 | - | 22 | 3661 | ||
AK126798.1 | - | 22 | 3573 | |||
AK304604.1 | - | 7 | 1106 | |||
BC039707.1 | - | 3 | 420 | 4479933 | ||
BC046145.1 | - | 4 | 512 | |||
KF175174.1 | - | 2 | 137 | |||
KF175210.1 | - | 3 | 380 | |||
KF175214.1 | - | 3 | 319 | |||
KF175219.1 | - | 2 | 287 |
Gene Symbol: PKD1P6-NPIPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_123721.1 | 20 | 4065 | |||
NR_123722.1 | 10 | 1926 | ||||
GenBank mRNA | AK131084.1 | - | 15 | 4429 | ||
AK131085.1 | - | 14 | 4335 | |||
AK302708.1 | - | 10 | 1926 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
