siRNA Details
Gene Symbol: CDC37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007065.3 | NP_008996.1 | 2 | 250 | ||
XM_011527652.2 | XP_011525954.1 | 2 | 217 | |||
GenBank mRNA | BC000083.2 | - | 2 | 174 | 3505011 | |
BC008793.2 | - | 2 | 174 | 3619769 | ||
BE796008.1 | - | 2 | 254 | |||
BT006796.1 | - | 2 | 134 | |||
CR542249.1 | - | 2 | 134 | |||
U43077.1 | - | 2 | 142 | |||
U63131.1 | - | 2 | 182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
