siRNA Details
Gene Symbol: MIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300867.1 | NP_001287796.1 | 7 | 777 | ||
NM_001324062.1 | NP_001310991.1 | 12 | 3979 | |||
NM_001324064.1 | NP_001310993.1 | 13 | 4024 | |||
NM_001324065.1 | NP_001310994.1 | 7 | 777 | |||
NM_198551.3 | NP_940953.2 | 12 | 3979 | |||
XM_011509513.2 | XP_011507815.2 | 12 | 4291 | |||
XM_017001243.1 | XP_016856732.1 | 12 | 3804 | |||
GenBank mRNA | AK025122.1 | - | 4 | 278 | ||
AK026489.1 | - | 10 | 985 | |||
AK290335.1 | - | 3 | 254 | |||
AK296712.1 | - | 7 | 758 | |||
AX775845.1 | - | 7 | 751 | |||
D87742.1 | - | 9 | 1828 | |||
L34688.1 | - | 1 | 57 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
