siRNA Details
Gene Symbol: MIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300867.1 | NP_001287796.1 | 9 | 948 | ||
NM_001324062.1 | NP_001310991.1 | 14 | 4150 | |||
NM_001324063.1 | NP_001310992.1 | 12 | 3973 | |||
NM_001324064.1 | NP_001310993.1 | 15 | 4195 | |||
NM_001324065.1 | NP_001310994.1 | 9 | 948 | |||
NM_198551.3 | NP_940953.2 | 14 | 4150 | |||
XM_006711304.3 | XP_006711367.2 | 12 | 4285 | |||
XM_011509513.2 | XP_011507815.2 | 14 | 4462 | |||
XM_017001243.1 | XP_016856732.1 | 14 | 3975 | |||
GenBank mRNA | AK025122.1 | - | 6 | 449 | ||
AK025506.1 | - | 11 | 1830 | |||
AK026489.1 | - | 12 | 1156 | |||
AK096526.1 | - | 9 | 2711 | |||
AK290335.1 | - | 5 | 425 | |||
AK296712.1 | - | 9 | 929 | |||
AX775845.1 | - | 9 | 922 | |||
D87742.1 | - | 11 | 1999 | |||
L34688.1 | - | 3 | 228 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
