siRNA Details
Gene Symbol: TMEM229B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182526.2 | NP_872332.1 | 3 | 490 | ||
XM_005267373.2 | XP_005267430.1 | 3 | 496 | |||
XM_005267374.3 | XP_005267431.1 | 4 | 363 | |||
XM_005267375.2 | XP_005267432.1 | 3 | 555 | |||
XM_006720062.2 | XP_006720125.1 | 3 | 375 | |||
XM_017021051.1 | XP_016876540.1 | 4 | 647 | |||
XM_017021052.1 | XP_016876541.1 | 3 | 718 | |||
XM_017021053.1 | XP_016876542.1 | 3 | 622 | |||
XM_017021054.1 | XP_016876543.1 | 3 | 860 | |||
XM_017021055.1 | XP_016876544.1 | 3 | 782 | |||
GenBank mRNA | AK090706.1 | - | 3 | 312 | ||
BC104940.1 | - | 2 | 226 | 8143943 | ||
BC104942.1 | - | 2 | 226 | 8143945 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
