siRNA Details
Gene Symbol: AJAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC041648.1 | - | 1 | 2022 |
Gene Symbol: BTBD18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017018127.1 | XP_016873616.1 | 1 | 687 |
Gene Symbol: DHX34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017027524.1 | XP_016883013.1 | 1 | 17 |
Gene Symbol: DSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001941.4 | NP_001932.2 | 16 | 4890 | ||
NM_024423.3 | NP_077741.2 | 17 | 4933 | |||
GenBank mRNA | AF297090.1 | - | 5 | 2993 | ||
AK074475.1 | - | 1 | 601 |
Gene Symbol: ERCC6L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020207.4 | NP_064592.2 | 19 | 7110 |
Gene Symbol: FBXO18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL832251.1 | - | 1 | 1304 |
Gene Symbol: G3BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK309172.1 | - | 4 | 1132 |
Gene Symbol: GLOD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF151908.1 | - | 1, 2 | 66 |
Gene Symbol: GNE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128227.2 | NP_001121699.1 | 12 | 4459 | ||
NM_001190383.1 | NP_001177312.1 | 11 | 4253 | |||
NM_001190384.1 | NP_001177313.1 | 10 | 4116 | |||
NM_001190388.1 | NP_001177317.1 | 11 | 4253 | |||
NM_005476.5 | NP_005467.1 | 12 | 4475 | |||
XM_017014167.1 | XP_016869656.1 | 12 | 4320 | |||
XM_017014168.1 | XP_016869657.1 | 11 | 4299 | |||
GenBank mRNA | AJ238764.1 | - | 12 | 2819 | ||
AY531126.1 | - | 12 | 2819 | |||
AY531127.1 | - | 12 | 2819 | |||
AY531128.1 | - | 12 | 2819 | |||
BQ217959.1 | - | 1 | 494 |
Gene Symbol: GOSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128816.1 | - | 1 | 1058 |
Gene Symbol: GPR34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK122945.1 | - | 1 | 1377 |
Gene Symbol: HMOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286270.1 | NP_001273199.1 | 3 | 456 | ||
GenBank mRNA | AK093388.1 | - | 3 | 417 |
Gene Symbol: ITK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209622.1 | - | 10 | 1915 |
Gene Symbol: KYAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR627392.1 | - | 10 | 2730 |
Gene Symbol: LOC100129721 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126643.1 | - | 1 | 2619 |
Gene Symbol: LOC100130027 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127225.1 | - | 5 | 3181 |
Gene Symbol: LONRF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005262476.2 | XP_005262533.1 | 12 | 5438 |
Gene Symbol: MGRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142289.2 | NP_001135761.2 | 16 | 2712 | ||
NM_001142290.2 | NP_001135762.1 | 17 | 5315 | |||
NM_001142291.2 | NP_001135763.2 | 16 | 5249 | |||
NM_015246.3 | NP_056061.1 | 17 | 2778 | |||
NR_102267.1 | 15 | 2602 | ||||
XM_005255217.1 | XP_005255274.1 | 17 | 2781 | |||
XM_005255218.1 | XP_005255275.1 | 16 | 2715 | |||
XM_011522441.1 | XP_011520743.1 | 17 | 2639 | |||
XM_011522442.2 | XP_011520744.1 | 9 | 2093 | |||
GenBank mRNA | AB011116.1 | - | 17 | 5295 | ||
BC007627.1 | - | 1 | 1899 | |||
BC050389.1 | - | 17 | 2748 | 5755971 |
Gene Symbol: MRPL45P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC090855.1 | - | 4 | 415 |
Gene Symbol: OPRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000914.4 | NP_000905.3 | 4 | 13146 | ||
NM_001145279.3 | NP_001138751.1 | 6 | 13505 | |||
NM_001145280.3 | NP_001138752.1 | 4 | 12936 | |||
NM_001145281.2 | NP_001138753.1 | 4 | 12477 | |||
NM_001145287.2 | NP_001138759.1 | 4 | 13711 | |||
NM_001285522.1 | NP_001272451.1 | 2 | 12272 | |||
NM_001285524.1 | NP_001272453.1 | 5 | 13041 | |||
NM_001285526.1 | NP_001272455.1 | 4 | 13692 | |||
NR_104348.1 | 5 | 12946 | ||||
NR_104350.1 | 3 | 12484 | ||||
XM_017010906.1 | XP_016866395.1 | 4 | 14621 |
Gene Symbol: P2RX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011538419.2 | XP_011536721.1 | 12 | 5665 | ||
XM_011538420.2 | XP_011536722.1 | 11 | 5332 | |||
XM_017019364.1 | XP_016874853.1 | 13 | 5978 | |||
XM_017019365.1 | XP_016874854.1 | 12 | 5706 | |||
XM_017019366.1 | XP_016874855.1 | 12 | 5693 | |||
XM_017019367.1 | XP_016874856.1 | 11 | 5531 |
Gene Symbol: PPP1R37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB075866.1 | - | 2 | 2030 |
Gene Symbol: PTPRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209333.1 | - | 30 | 4743 |
Gene Symbol: RPP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006413.4 | NP_006404.1 | 11 | 2034 | ||
GenBank mRNA | AL833628.1 | - | 11 | 2022 | ||
BC006991.1 | - | 12 | 2296 | 3827681 |
Gene Symbol: SERPINB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004155.5 | NP_004146.1 | 7 | 1756 | ||
XM_005249184.4 | XP_005249241.1 | 7 | 1919 | |||
XM_011514678.2 | XP_011512980.1 | 7 | 1767 | |||
XM_017010942.1 | XP_016866431.1 | 8 | 1906 | |||
XM_017010943.1 | XP_016866432.1 | 8 | 2064 | |||
GenBank mRNA | AB060690.1 | - | 7 | 1718 | ||
BC002538.2 | - | 7 | 1717 | 3140427 | ||
BX640606.1 | - | 2 | 1235 |
Gene Symbol: TESK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK299269.1 | - | 4 | 1211 |
Gene Symbol: TM2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK311444.1 | - | 6 | 1150 |
Gene Symbol: TRDMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321006.1 | NP_001307935.1 | 9 | 12715 | ||
NM_001321007.1 | NP_001307936.1 | 12 | 13088 | |||
NM_004412.6 | NP_004403.1 | 11 | 12858 | |||
XM_005252374.4 | XP_005252431.1 | 11 | 2914 |
Gene Symbol: UHMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184763.1 | NP_001171692.1 | 8 | 3512 | ||
NM_144624.2 | NP_653225.2 | 7 | 3764 | |||
NM_175866.4 | NP_787062.1 | 8 | 3853 |
Gene Symbol: VPS28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU658837.1 | - | 2 | 84 |
Gene Symbol: XIAP-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM931494.1 | - | 1 | 35 |
Gene Symbol: ZBTB16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126422.1 | - | 1 | 827 |
Gene Symbol: ZFX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005274594.4 | XP_005274651.1 | 1 | 655 |
Gene Symbol: ZFYVE21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK226142.1 | - | 1 | 2503 |
Gene Symbol: ZMYM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289091.1 | NP_001276020.1 | 2 | 114 | ||
GenBank mRNA | AK096206.1 | - | 2 | 91 | ||
AK308655.1 | - | 2 | 91 |
Gene Symbol: ZNF114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC125068.1 | - | 122 | 40117544 |
Gene Symbol: ZNF135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095796.1 | - | 3 | 457 |
Gene Symbol: ZNF517 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA782312.1 | - | 1 | 267 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
