siRNA Details
Gene Symbol: SPANXA2-OT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037183.2 | 1, 2 | 401 | |||
GenBank mRNA | AK093505.1 | - | 1, 2 | 416 | ||
DA220378.1 | - | 1, 2 | 119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
