siRNA Details
Gene Symbol: DCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000555.3 | NP_000546.2 | 7 | 6624 | ||
NM_001195553.1 | NP_001182482.1 | 7 | 6344 | |||
NM_178151.2 | NP_835364.1 | 7 | 6453 | |||
NM_178152.2 | NP_835365.1 | 7 | 6341 | |||
NM_178153.2 | NP_835366.1 | 7 | 6326 | |||
XM_011530878.2 | XP_011529180.1 | 7 | 6301 | |||
XM_011530879.2 | XP_011529181.1 | 6 | 6218 | |||
XM_011530880.2 | XP_011529182.1 | 6 | 6203 | |||
XM_017029312.1 | XP_016884801.1 | 7 | 6409 | |||
GenBank mRNA | AJ003112.1 | - | 7 | 6634 |
Gene Symbol: CDKN2B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003529.3 | 7 | 1280 | |||
NR_047532.1 | 6 | 967 | ||||
NR_047535.1 | 4 | 672 | ||||
NR_047537.1 | 4 | 672 | ||||
NR_047539.1 | 7 | 1280 | ||||
NR_047540.1 | 4 | 672 | ||||
NR_047541.1 | 4 | 672 | ||||
NR_047542.1 | 4 | 672 | ||||
NR_047543.1 | 4 | 672 | ||||
GenBank mRNA | AB548314.1 | - | 6 | 967 | ||
DQ485453.1 | - | 7 | 1279 | |||
DQ485454.1 | - | 7 | 1279 | |||
EU741058.1 | - | 4 | 368 | |||
GQ495921.1 | - | 4 | 561 | |||
GQ495922.1 | - | 4 | 561 | |||
GQ495923.1 | - | 4 | 561 | |||
GQ495924.1 | - | 4 | 561 | |||
GQ495926.1 | - | 4 | 672 |
Gene Symbol: CYCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018947.5 | NP_061820.1 | 3 | 2838 | ||
GenBank mRNA | AK056360.1 | - | 1 | 236 | ||
AL713681.1 | - | 3 | 2728 |
Gene Symbol: DDX55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017019714.1 | XP_016875203.1 | 2 | 327 |
Gene Symbol: LAMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014398.3 | NP_055213.2 | 6 | 2189 | ||
GenBank mRNA | AB013924.1 | - | 6 | 2027 | ||
AJ005766.1 | - | 6 | 2005 | |||
BC032940.2 | - | 6 | 2145 | 4829125 |
Gene Symbol: LINC00596 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX161431.1 | - | 1 | 387 |
Gene Symbol: LINC01102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055090.1 | - | 2 | 971 | ||
AK126958.1 | - | 2 | 2060 |
Gene Symbol: LINC01562 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127900.1 | - | 2 | 461 |
Gene Symbol: MZF1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027334.2 | 6 | 2431 |
Gene Symbol: PHF20L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX641040.1 | - | 1 | 3908 |
Gene Symbol: RRN3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126166.1 | - | 1 | 201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
