siRNA Details
Gene Symbol: ANXA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040084.2 | NP_001035173.1 | 11 | 1105 | ||
NM_001271702.1 | NP_001258631.1 | 11 | 1219 | |||
NM_001271703.1 | NP_001258632.1 | 9 | 919 | |||
XM_006717951.3 | XP_006718014.1 | 11 | 1059 | |||
XM_011540096.1 | XP_011538398.1 | 12 | 1258 | |||
XM_011540098.1 | XP_011538400.1 | 11 | 1162 | |||
GenBank mRNA | AK296573.1 | - | 9 | 770 | ||
AK298761.1 | - | 11 | 1085 | |||
AK300139.1 | - | 11 | 1070 | |||
AK300158.1 | - | 10 | 883 | |||
BC004376.1 | - | 11 | 1002 | 3639064 | ||
BC073755.1 | - | 11 | 1036 | 6538697 | ||
M81844.1 | - | 11 | 907 |
Gene Symbol: ANXA8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098845.2 | NP_001092315.2 | 11 | 1142 | ||
NM_001278923.1 | NP_001265852.1 | 9 | 971 | |||
NM_001278924.1 | NP_001265853.1 | 8 | 989 | |||
GenBank mRNA | AK223149.1 | - | 11 | 956 | ||
AK300191.1 | - | 9 | 805 | |||
AK313626.1 | - | 11 | 1005 | |||
BC008813.2 | - | 8 | 903 | 3958020 | ||
X16662.1 | - | 11 | 986 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
