siRNA Details
Gene Symbol: ANXA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040084.2 | NP_001035173.1 | 11, 12 | 1153 | ||
NM_001271702.1 | NP_001258631.1 | 11, 12 | 1267 | |||
NM_001271703.1 | NP_001258632.1 | 9, 10 | 967 | |||
XM_006717951.3 | XP_006718014.1 | 11, 12 | 1107 | |||
XM_011540096.1 | XP_011538398.1 | 12, 13 | 1306 | |||
XM_011540098.1 | XP_011538400.1 | 11, 12 | 1210 | |||
GenBank mRNA | AK296573.1 | - | 9, 10 | 818 | ||
AK298761.1 | - | 11, 12 | 1133 | |||
AK300139.1 | - | 11, 12 | 1118 | |||
AK300158.1 | - | 10, 11 | 931 | |||
BC004376.1 | - | 11, 12 | 1050 | 3639064 | ||
BC073755.1 | - | 11, 12 | 1084 | 6538697 |
Gene Symbol: ANXA8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098845.2 | NP_001092315.2 | 11, 12 | 1190 | ||
NM_001278923.1 | NP_001265852.1 | 9, 10 | 1019 | |||
NM_001278924.1 | NP_001265853.1 | 8, 9 | 1037 | |||
GenBank mRNA | AK223149.1 | - | 11, 12 | 1004 | ||
AK300191.1 | - | 9, 10 | 853 | |||
AK313626.1 | - | 11, 12 | 1053 | |||
BC008813.2 | - | 8, 9 | 951 | 3958020 | ||
X16662.1 | - | 11, 12 | 1034 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
