siRNA Details
Gene Symbol: FBXL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005266930.2 | XP_005266987.1 | 9 | 5529 | ||
XM_017010726.1 | XP_016866215.1 | 9 | 5427 | |||
XM_017010727.1 | XP_016866216.1 | 8 | 5383 | |||
XM_017010728.1 | XP_016866217.1 | 11 | 5689 | |||
GenBank mRNA | AK055220.1 | - | 1 | 149 |
Gene Symbol: ALG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019109.4 | NP_061982.3 | 13 | 3285 |
Gene Symbol: ASB16-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_049729.1 | 2 | 192 | |||
GenBank mRNA | AK055685.1 | - | 2 | 146 | ||
CB151608.1 | - | 2 | 189 |
Gene Symbol: C16orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175900.3 | NP_787096.2 | 2 | 1854 | ||
GenBank mRNA | AK093000.1 | - | 2 | 1841 |
Gene Symbol: CLSTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017007022.1 | XP_016862511.1 | 17 | 6584 |
Gene Symbol: COMMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM981499.1 | - | 1 | 397 |
Gene Symbol: CTRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011540550.1 | XP_011538852.1 | 7 | 2327 | ||
GenBank mRNA | CR749296.1 | - | 5 | 2134 |
Gene Symbol: DHRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126383.1 | - | 1 | 1128 |
Gene Symbol: DLEU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002612.1 | 11 | 2157 | |||
GenBank mRNA | AF529010.1 | - | 4 | 1175 | ||
AY455757.1 | - | LIT3420 DLEU2 |
14 | 2457 |
Gene Symbol: DLEU7-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046551.1 | 5 | 984 | |||
GenBank mRNA | BC035769.1 | - | 5 | 994 |
Gene Symbol: FAM35A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK025471.1 | - | 1 | 810 |
Gene Symbol: FZD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017412.3 | NP_059108.1 | 8 | 6732 | ||
NM_145866.1 | NP_665873.1 | 7 | 6686 |
Gene Symbol: GAS2L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303130.1 | NP_001290059.1 | 9 | 4684 | ||
NM_001303131.1 | NP_001290060.1 | 10 | 5171 | |||
NM_174942.2 | NP_777602.1 | 10 | 4805 |
Gene Symbol: GJC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR933719.1 | - | 3 | 1754 |
Gene Symbol: HOOK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032410.3 | NP_115786.1 | 22 | 10852 |
Gene Symbol: ISPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101417.3 | NP_001094887.1 | 9 | 2220 | ||
NM_001101426.3 | NP_001094896.1 | 10 | 2370 |
Gene Symbol: KIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK309059.1 | - | 8 | 1069 |
Gene Symbol: LGI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017016911.1 | XP_016872400.1 | 8 | 1292 |
Gene Symbol: LINC01205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109842.1 | 2 | 2365 | |||
GenBank mRNA | BC044821.1 | - | 2 | 2196 |
Gene Symbol: LINC01569 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039999.1 | 2 | 526 | |||
GenBank mRNA | AK094828.1 | - | 2 | 526 |
Gene Symbol: LNX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153371.3 | NP_699202.1 | 10 | 2903 | ||
XM_011534995.2 | XP_011533297.1 | 9 | 2534 | |||
XM_017020434.1 | XP_016875923.1 | 10 | 2762 | |||
GenBank mRNA | AK022995.1 | - | 2 | 676 | ||
BC036755.2 | - | 10 | 2732 | 5541168 |
Gene Symbol: LOC100133306 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125136.1 | - | 1 | 1442 |
Gene Symbol: LOC101929224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110787.1 | 2 | 155 | |||
GenBank mRNA | BC042016.1 | - | 2 | 155 |
Gene Symbol: LOC105371451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125505.1 | - | 1 | 1204 |
Gene Symbol: LOC105371998 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090999.1 | - | 1 | 814 |
Gene Symbol: LOC400682 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011528477.2 | XP_011526779.2 | 4 | 2835 | ||
GenBank mRNA | BX647610.1 | - | 2 | 2637 |
Gene Symbol: NMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC005232.1 | - | 2 | 661 |
Gene Symbol: NT5DC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031701.2 | NP_001026871.1 | 14 | 3004 | ||
GenBank mRNA | AB032773.1 | - | 14 | 2802 |
Gene Symbol: PDCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017004890.1 | XP_016860379.1 | 6 | 1606 |
Gene Symbol: PIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092970.1 | - | 1 | 1465 |
Gene Symbol: PPP2R3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC037797.1 | - | 1 | 1429 |
Gene Symbol: PTPRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109754.2 | NP_001103224.1 | 34 | 9794 | ||
NM_001206971.1 | NP_001193900.1 | 31 | 8855 | |||
NM_001206972.1 | NP_001193901.1 | 31 | 8855 | |||
NM_002837.4 | NP_002828.3 | 32 | 9125 | |||
GenBank mRNA | AK023720.1 | - | 1 | 307 | ||
AK097371.1 | - | 1 | 1627 |
Gene Symbol: RNF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010077.1 | XP_016865566.1 | 5 | 3655 |
Gene Symbol: RPLP0P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002775.2 | 5 | 2847 | |||
GenBank mRNA | AK095332.1 | - | 5 | 2850 |
Gene Symbol: RTCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130841.1 | NP_001124313.1 | 12 | 2216 | ||
NM_003729.3 | NP_003720.1 | 11 | 2177 | |||
XM_005271298.4 | XP_005271355.1 | 11 | 2174 | |||
XM_005271299.4 | XP_005271356.1 | 11 | 2163 | |||
XM_017002669.1 | XP_016858158.1 | 11 | 1920 | |||
GenBank mRNA | CA437545.1 | - | 1 | 433 | ||
DB576272.1 | - | 1 | 89 |
Gene Symbol: SPATS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG370341.1 | - | 1 | 248 | ||
DB305045.1 | - | 1 | 240 |
Gene Symbol: TBRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092132.1 | - | 1 | 731 |
Gene Symbol: TSIX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003255.2 | 1 | 18429 |
Gene Symbol: TUBGCP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017021890.1 | XP_016877379.1 | 23 | 3118 | ||
XM_017021891.1 | XP_016877380.1 | 23 | 3115 | |||
XM_017021894.1 | XP_016877383.1 | 20 | 2766 | |||
XM_017021895.1 | XP_016877384.1 | 23 | 3135 | |||
XM_017021896.1 | XP_016877385.1 | 15 | 2250 | |||
XM_017021897.1 | XP_016877386.1 | 14 | 2010 | |||
XM_017021898.1 | XP_016877387.1 | 18 | 2571 | |||
XM_017021899.1 | XP_016877388.1 | 23 | 3156 | |||
XM_017021900.1 | XP_016877389.1 | 22 | 3031 | |||
XM_017021901.1 | XP_016877390.1 | 14 | 3080 | |||
GenBank mRNA | AK310897.1 | - | 24 | 3222 |
Gene Symbol: UGGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020120.3 | NP_064505.1 | 41 | 9430 | ||
NR_027671.2 | 41 | 9583 | ||||
XM_006712634.3 | XP_006712697.1 | 41 | 9404 | |||
XM_006712635.3 | XP_006712698.1 | 40 | 9374 | |||
XM_006712636.3 | XP_006712699.1 | 41 | 9393 | |||
XM_017004507.1 | XP_016859996.1 | 42 | 9489 | |||
XM_017004508.1 | XP_016859997.1 | 42 | 9444 | |||
XM_017004509.1 | XP_016859998.1 | 27 | 7735 | |||
XM_017004510.1 | XP_016859999.1 | 23 | 8114 |
Gene Symbol: USP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X78261.1 | - | 440 |
Gene Symbol: WBP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK308843.1 | - | 4 | 463 |
Gene Symbol: WDR45B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017024862.1 | XP_016880351.1 | 5 | 960 |
Gene Symbol: ZFAND2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012774.1 | XP_016868263.1 | 6 | 1363 |
Gene Symbol: ZNF454 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323307.1 | NP_001310236.1 | 5 | 3371 | ||
NM_001323308.1 | NP_001310237.1 | 5 | 3368 | |||
NM_001323309.1 | NP_001310238.1 | 5 | 3310 | |||
NM_001323310.1 | NP_001310239.1 | 5 | 3313 |
Gene Symbol: ZNF708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297560.1 | NP_001284489.1 | 4 | 3425 | ||
NM_001297561.1 | NP_001284490.1 | 3 | 3216 | |||
NM_021269.2 | NP_067092.2 | 4 | 3341 | |||
XM_017027202.1 | XP_016882691.1 | 4 | 3366 | |||
XM_017027203.1 | XP_016882692.1 | 2 | 3146 | |||
XM_017027204.1 | XP_016882693.1 | 5 | 2907 | |||
GenBank mRNA | AK128184.1 | - | 5 | 3167 |
Gene Symbol: ZNF891 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017018666.1 | XP_016874155.1 | 2 | 2939 | ||
XM_017018667.1 | XP_016874156.1 | 2 | 2609 | |||
XM_017018668.1 | XP_016874157.1 | 2 | 2692 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
