siRNA Details
Gene Symbol: CTAGE10P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003268.3 | 1 | 1320 | |||
GenBank mRNA | BC030655.1 | - | 1 | 1219 |
Gene Symbol: CTAGE11P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027466.1 | 1 | 1170 | |||
GenBank mRNA | AL133018.1 | - | 1 | 1170 |
Gene Symbol: CTAGE15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008747.2 | NP_001008747.1 | 1 | 1161 | ||
GenBank mRNA | BC146818.1 | - | 1 | 1161 | 8860423 |
Gene Symbol: CTAGE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198495.2 | NP_940897.2 | 1 | 1193 | ||
GenBank mRNA | AF338232.1 | - | 1 | 522 |
Gene Symbol: CTAGE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178561.4 | NP_848656.2 | 1 | 1236 | ||
GenBank mRNA | BC101323.1 | - | 1 | 494 | ||
BC101324.2 | - | 1 | 494 |
Gene Symbol: CTAGE7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_044994.1 | 1 | 1169 | |||
GenBank mRNA | BC036527.1 | - | 1 | 1172 |
Gene Symbol: CTAGE8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278507.1 | NP_001265436.1 | 1 | 1678 | ||
GenBank mRNA | AK292236.1 | - | 1 | 1182 |
Gene Symbol: CTAGE9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145659.1 | NP_001139131.1 | 1 | 1144 | ||
GenBank mRNA | BC101322.1 | - | 1 | 494 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
