siRNA Details
Gene Symbol: KCNQ1OT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002728.3 | 1 | 63772 |
Gene Symbol: HELLPAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | JX088243.1 | - | 1 | 41786 |
Gene Symbol: LINC00504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126435.1 | 7 | 1905 | |||
GenBank mRNA | BC036758.2 | - | 7 | 1917 | 5563071 |
Gene Symbol: LOC100133091 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC090058.1 | - | 2 | 1786 |
Gene Symbol: MIPOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647795.1 | - | 10 | 5372 |
Gene Symbol: TBC1D8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648230.1 | - | 7 | 2928 |
Gene Symbol: UGDH-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047679.1 | 5 | 1784 | |||
GenBank mRNA | AK056558.1 | - | 5 | 1604 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
