siRNA Details
Gene Symbol: IGHG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128301.1 | - | 2 | 158 | ||
BC006402.1 | - | 2 | 195 | |||
BC019046.2 | - | 2 | 184 | |||
BC069020.1 | - | 2 | 167 | |||
BC092518.1 | - | 2 | 181 | |||
BX640622.1 | - | 2 | 200 | |||
BX640627.1 | - | 2 | 201 |
Gene Symbol: IGHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF067420.1 | - | 2 | 189 | ||
AK027379.1 | - | 2 | 197 | |||
AK128476.1 | - | 2 | 200 | |||
AK128565.1 | - | 2 | 201 | |||
BC073771.1 | - | 2 | 177 |
Gene Symbol: IGHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128658.1 | - | 2 | 201 | ||
BC021276.2 | - | 2 | 169 |
Gene Symbol: IGHG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC062335.1 | - | 2 | 184 |
Gene Symbol: IGHM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC015760.1 | - | 2 | 180 | ||
BC020240.1 | - | 2 | 191 | |||
BC041037.1 | - | 2 | 169 | |||
BC073758.1 | - | 2 | 190 |
Gene Symbol: IGHV3-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF231396.1 | - | 1 | 64 | ||
AJ519334.1 | - | 1 | 94 | |||
X14584.1 | - | 2 | 145 |
Gene Symbol: IGHV3-41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HM774330.1 | - | 41 |
Gene Symbol: IGHV3-48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF035023.1 | - | 1 | 64 |
Gene Symbol: IGHV4-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY172959.1 | - | 2 | 121 | ||
BX640947.1 | - | 2 | 201 |
Gene Symbol: IGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF103353.1 | - | 39 | |||
AJ239369.1 | - | 1 | 64 |
Gene Symbol: IGKV3-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ006111.1 | - | 64 | |||
AJ006112.1 | - | 64 | ||||
AJ006114.1 | - | 64 | ||||
AJ006115.1 | - | 64 | ||||
AJ006117.1 | - | 64 | ||||
AJ006118.1 | - | 64 | ||||
AJ006119.1 | - | 64 | ||||
AJ006120.1 | - | 64 |
Gene Symbol: IGLJ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB001733.1 | - | 67 | |||
AB001736.1 | - | 67 |
Gene Symbol: LAIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | KU058441.1 | - | 64 |
Gene Symbol: LOC105379528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030112.1 | XP_016885601.1 | 2 | 1249 |
Gene Symbol: SCFV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | Y13056.1 | - | 64 |
Gene Symbol: ZCWPW2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB087877.1 | - | 1 | 78 | ||
AJ519236.1 | - | 1 | 94 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
