siRNA Details
Gene Symbol: HOMER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145721.1 | NP_001139193.1 | 3 | 214 | ||
NM_001145722.1 | NP_001139194.1 | 3 | 636 | |||
NM_001145724.1 | NP_001139196.1 | 2 | 182 | |||
NM_004838.3 | NP_004829.3 | 3 | 364 | |||
NR_027297.1 | 2 | 768 | ||||
XM_006722943.1 | XP_006723006.1 | 3 | 248 | |||
XM_006722944.1 | XP_006723007.1 | 3 | 266 | |||
GenBank mRNA | AF093265.1 | - | 3 | 205 | ||
AK124450.1 | - | 2 | 768 | |||
AK313995.1 | - | 3 | 637 | |||
AL529052.3 | - | 3 | 323 | |||
BC012113.1 | - | 3 | 306 | 4555641 | ||
BT019464.1 | - | 2 | 115 | |||
Y17573.1 | - | 2 | 115 | |||
Y18894.1 | - | 2 | 115 | |||
Y18895.1 | - | 2 | 115 | |||
Y18896.1 | - | 2 | 115 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
