siRNA Details
Gene Symbol: HOMER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145721.1 | NP_001139193.1 | 4 | 286 | ||
NM_001145722.1 | NP_001139194.1 | 4 | 708 | |||
NM_001145724.1 | NP_001139196.1 | 3 | 254 | |||
NM_004838.3 | NP_004829.3 | 4 | 436 | |||
NR_027297.1 | 3 | 840 | ||||
XM_006722943.1 | XP_006723006.1 | 4 | 320 | |||
XM_006722944.1 | XP_006723007.1 | 4 | 338 | |||
GenBank mRNA | AF093265.1 | - | 4 | 277 | ||
AK124450.1 | - | 3 | 840 | |||
AK313995.1 | - | 4 | 709 | |||
AL529052.3 | - | 4 | 395 | |||
BC012113.1 | - | 4 | 378 | 4555641 | ||
BT019464.1 | - | 3 | 187 | |||
Y17573.1 | - | 3 | 187 | |||
Y18894.1 | - | 3 | 187 | |||
Y18895.1 | - | 3 | 187 | |||
Y18896.1 | - | 3 | 187 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
