siRNA Details
Gene Symbol: CYSLTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK023929.1 | - | 2 | 1175 |
Gene Symbol: ATP9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK295981.1 | - | 5 | 858 | ||
BC053561.1 | - | 5 | 776 |
Gene Symbol: KCNQ1OT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002728.3 | 1 | 64173 |
Gene Symbol: LOC101929163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097727.1 | - | 1 | 2105 | ||
AK123889.1 | - | 2 | 2905 |
Gene Symbol: NEK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321221.1 | NP_001308150.1 | 5 | 729 | ||
XM_017007205.1 | XP_016862694.1 | 5 | 906 | |||
XM_017007206.1 | XP_016862695.1 | 5 | 824 | |||
XM_017007207.1 | XP_016862696.1 | 5 | 1040 | |||
XM_017007208.1 | XP_016862697.1 | 4 | 624 | |||
XM_017007209.1 | XP_016862698.1 | 4 | 833 | |||
XM_017007210.1 | XP_016862699.1 | 4 | 689 | |||
XM_017007211.1 | XP_016862700.1 | 4 | 624 | |||
XM_017007218.1 | XP_016862707.1 | 6 | 807 | |||
XM_017007219.1 | XP_016862708.1 | 3 | 301 | |||
XM_017007220.1 | XP_016862709.1 | 4 | 624 | |||
XM_017007221.1 | XP_016862710.1 | 4 | 624 | |||
XM_017007222.1 | XP_016862711.1 | 4 | 624 |
Gene Symbol: RABGAP1L-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX485272.1 | - | 2 | 615 |
Gene Symbol: RIC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006718321.3 | XP_006718384.1 | 5 | 839 |
Gene Symbol: SPAG16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017004899.1 | XP_016860388.1 | 9 | 1008 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
