siRNA Details
Gene Symbol: BMS1P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026592.2 | 2 | 123 | |||
GenBank mRNA | AK294783.1 | - | 2 | 123 | ||
BX955228.1 | - | 2 | 112 |
Gene Symbol: BMS1P21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033857.1 | 2 | 199 | |||
GenBank mRNA | AK307851.1 | - | 2 | 191 |
Gene Symbol: BMS1P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003611.2 | 2 | 210 | |||
GenBank mRNA | AK307333.1 | - | 2 | 69 | ||
AL832069.1 | - | 2 | 97 | |||
BC017177.2 | - | 1 | 58 | |||
BC065722.1 | - | 2 | 79 | |||
BC131568.1 | - | 1 | 864 | |||
DA245014.1 | - | 2 | 103 |
Gene Symbol: BMS1P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024495.2 | 2 | 92 | |||
GenBank mRNA | BC051298.1 | - | 1 | 58 | ||
BG474237.1 | - | 1 | 59 | |||
CD684353.1 | - | 2 | 116 | |||
CN417542.1 | - | 1 | 53 | |||
DA115686.1 | - | 2 | 75 | |||
DB072344.1 | - | 2 | 92 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
