siRNA Details
Gene Symbol: BMS1P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026592.2 | 3 | 300 | |||
GenBank mRNA | AK294783.1 | - | 3 | 300 | ||
BX955228.1 | - | 3 | 289 |
Gene Symbol: AGAP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133447.1 | NP_597704.1 | 5 | 602 | ||
GenBank mRNA | AB075855.1 | - | 2 | 79 | ||
AK097165.1 | - | 5 | 602 |
Gene Symbol: BMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014753.3 | NP_055568.3 | 3 | 549 | ||
XM_005271846.3 | XP_005271903.1 | 3 | 424 | |||
XM_005271848.3 | XP_005271905.1 | 3 | 280 | |||
XM_005271849.3 | XP_005271906.1 | 3 | 280 | |||
XM_011540402.2 | XP_011538704.1 | 3 | 280 | |||
GenBank mRNA | AK308420.1 | - | 3 | 256 | ||
BC043345.1 | - | 3 | 244 | 6062924 | ||
BC150252.1 | - | 2 | 218 | 9007274 | ||
D80009.1 | - | 2 | 218 | |||
DB064246.1 | - | 3 | 518 |
Gene Symbol: BMS1P21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033857.1 | 3 | 376 | |||
GenBank mRNA | AK307851.1 | - | 3 | 368 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
