siRNA Details
Gene Symbol: ERVK-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ069911.1 | - | 1 | 1045 | ||
DQ069912.1 | - | 1 | 1045 | |||
DQ069913.1 | - | 1 | 1048 | |||
DQ069914.1 | - | 1 | 1045 | |||
DQ069915.1 | - | 1 | 1045 | |||
DQ069916.1 | - | 1 | 1045 | |||
DQ069917.1 | - | 1 | 1045 | |||
DQ069918.1 | - | 1 | 1045 |
Gene Symbol: ERVK-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AU124350.1 | - | 1 | 200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
