siRNA Details
Gene Symbol: SLC7A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003486.5 | NP_003477.4 | 1 | 131 | ||
XM_017023735.1 | XP_016879224.1 | 1 | 136 | |||
XM_017023736.1 | XP_016879225.1 | 1 | 136 | |||
GenBank mRNA | AB017908.1 | - | 1 | 99 | ||
AB018009.1 | - | 1 | 124 | |||
AB018542.1 | - | 1 | 74 | |||
AF077866.1 | - | 1 | 111 | |||
AF104032.1 | - | 1 | 125 | |||
BC039692.2 | - | 1 | 156 | 5551612 | ||
BC042600.1 | - | 1 | 125 | 4470593 |
Gene Symbol: SLC7A5P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002594.1 | 1 | 138 | |||
GenBank mRNA | BC071656.1 | - | 1 | 111 | ||
BM904244.1 | - | 110 |
Gene Symbol: SMG1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135317.1 | 1 | 136 | |||
GenBank mRNA | AB055226.1 | - | 1 | 138 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
