siRNA Details
Gene Symbol: MIGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017001231.1 | XP_016856720.1 | 15 | 1652 | ||
XM_017001232.1 | XP_016856721.1 | 15 | 1710 |
Gene Symbol: ALDH1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056534.1 | - | 3 | 261 |
Gene Symbol: DDA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024050.5 | NP_076955.1 | 5 | 1995 |
Gene Symbol: LINC01012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038292.1 | 3 | 2184 | |||
GenBank mRNA | BC035101.1 | - | 3 | 2185 |
Gene Symbol: LOC100128400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124238.1 | - | 1 | 1321 |
Gene Symbol: PSMA3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055868.1 | - | 2 | 221 |
Gene Symbol: QTRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055216.1 | - | 1 | 506 |
Gene Symbol: RBMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017019777.1 | XP_016875266.1 | 4 | 398 | ||
GenBank mRNA | AK293315.1 | - | 4 | 282 |
Gene Symbol: SLC35F6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017877.3 | NP_060347.2 | 6 | 1705 | ||
GenBank mRNA | BX647807.1 | - | 5 | 1623 | ||
BX648017.1 | - | 5 | 1483 |
Gene Symbol: SRRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011530178.2 | XP_011528480.1 | 7 | 2529 |
Gene Symbol: TADA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006722044.3 | XP_006722107.1 | 16 | 2188 | ||
XM_011525153.2 | XP_011523455.1 | 15 | 1961 | |||
XM_011525154.2 | XP_011523456.1 | 15 | 2180 | |||
XM_011525155.2 | XP_011523457.1 | 14 | 1869 | |||
XM_017024981.1 | XP_016880470.1 | 15 | 1995 | |||
XM_017024982.1 | XP_016880471.1 | 14 | 1897 | |||
XM_017024983.1 | XP_016880472.1 | 16 | 2152 | |||
XM_017024984.1 | XP_016880473.1 | 15 | 1976 | |||
XM_017024985.1 | XP_016880474.1 | 15 | 2137 | |||
XM_017024986.1 | XP_016880475.1 | 15 | 1970 |
Gene Symbol: THAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_133920.1 | 4 | 551 | |||
XM_005262774.3 | XP_005262831.1 | 6 | 2655 | |||
GenBank mRNA | AK056959.1 | - | 4 | 511 | ||
AK308914.1 | - | 6 | 672 |
Gene Symbol: UNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006722145.3 | XP_006722208.1 | 4 | 4449 | ||
XM_017025247.1 | XP_016880736.1 | 4 | 4449 | |||
XM_017025248.1 | XP_016880737.1 | 3 | 4351 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
