siRNA Details
Gene Symbol: SP100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080391.1 | NP_001073860.1 | 29 | 3795 | ||
GenBank mRNA | AK090565.1 | - | 1 | 1002 | ||
AK160379.1 | - | 5 | 3186 | |||
CR749288.1 | - | 8 | 1774 |
Gene Symbol: ATP9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC053561.1 | - | 5 | 1040 |
Gene Symbol: CFAP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC041426.1 | - | 7 | 1363 |
Gene Symbol: KCNQ1OT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002728.3 | 1 | 64438 |
Gene Symbol: LINC00504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126435.1 | 7 | 2569 | |||
GenBank mRNA | BC036758.2 | - | 7 | 2581 | 5563071 |
Gene Symbol: LOC100129434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125368.1 | 3 | 2345 | |||
GenBank mRNA | AK096196.1 | - | 3 | 2554 |
Gene Symbol: LOC100131257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034022.1 | 1 | 11973 |
Gene Symbol: LOC101926898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125837.1 | 3 | 823 | |||
GenBank mRNA | BC042616.1 | - | 3 | 825 |
Gene Symbol: LOC101927374 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110133.1 | 3 | 862 | |||
GenBank mRNA | AK094694.1 | - | 3 | 862 |
Gene Symbol: LOC101929163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123889.1 | - | 2 | 3169 |
Gene Symbol: MIPOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647795.1 | - | 10 | 6032 |
Gene Symbol: RIC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006718321.3 | XP_006718384.1 | 5 | 1103 |
Gene Symbol: ROR1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110665.1 | 4 | 1585 | |||
GenBank mRNA | AK096291.1 | - | 5 | 1675 |
Gene Symbol: SMC5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR936772.1 | - | 3 | 6227 |
Gene Symbol: TBC1D8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648230.1 | - | 7 | 3592 |
Gene Symbol: TMEM212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164436.1 | NP_001157908.1 | 5 | 768 | ||
GenBank mRNA | AK026825.1 | - | 5 | 748 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
