siRNA Details
Gene Symbol: KIF22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256269.1 | NP_001243198.1 | 13 | 1853 | ||
NM_001256270.1 | NP_001243199.1 | 12 | 1941 | |||
NM_007317.2 | NP_015556.1 | 12 | 1804 | |||
GenBank mRNA | AB017430.2 | - | 12 | 1781 | ||
AK223431.1 | - | 12 | 1790 | |||
AK294380.1 | - | 12 | 2180 | |||
AK312234.1 | - | 12 | 1795 | |||
AK316050.1 | - | 12 | 1941 | |||
AK316389.1 | - | 13 | 1812 | |||
BC004352.1 | - | 12 | 1776 | 3535435 | ||
BC028155.1 | - | 12 | 1762 | 5241557 | ||
BT007259.1 | - | 12 | 1757 | |||
L29095.1 | - | 1 | 94 | |||
L29096.1 | - | 3 | 274 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
