siRNA Details
Gene Symbol: KIF22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256269.1 | NP_001243198.1 | 12 | 1747 | ||
NM_001256270.1 | NP_001243199.1 | 11 | 1835 | |||
NM_007317.2 | NP_015556.1 | 11 | 1698 | |||
GenBank mRNA | AB017430.2 | - | 11 | 1675 | ||
AK223431.1 | - | 11 | 1684 | |||
AK312234.1 | - | 11 | 1689 | |||
AK316050.1 | - | 11 | 1835 | |||
AK316389.1 | - | 12 | 1706 | |||
BC004352.1 | - | 11 | 1670 | 3535435 | ||
BC028155.1 | - | 11 | 1656 | 5241557 | ||
BT007259.1 | - | 11 | 1651 | |||
L29096.1 | - | 2 | 168 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
