siRNA Details
Gene Symbol: KIF22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256269.1 | NP_001243198.1 | 15 | 2117 | ||
NM_001256270.1 | NP_001243199.1 | 14 | 2205 | |||
NM_007317.2 | NP_015556.1 | 14 | 2068 | |||
GenBank mRNA | AB017430.2 | - | 14 | 2045 | ||
AK223431.1 | - | 14 | 2054 | |||
AK294380.1 | - | 14 | 2444 | |||
AK316050.1 | - | 14 | 2205 | |||
AK316389.1 | - | 15 | 2076 | |||
BC004352.1 | - | 14 | 2040 | 3535435 | ||
BC028155.1 | - | 14 | 2026 | 5241557 | ||
L29095.1 | - | 3 | 358 | |||
L29096.1 | - | 5 | 538 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
