siRNA Details
Gene Symbol: FAHD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320848.1 | NP_001307777.1 | 7 | 1086 | ||
NM_199336.2 | NP_955368.1 | 6 | 960 | |||
XM_011510745.2 | XP_011509047.1 | 7 | 1076 | |||
XM_011510746.2 | XP_011509048.1 | 7 | 1195 | |||
XM_011510747.2 | XP_011509049.1 | 6 | 960 | |||
XM_011510748.2 | XP_011509050.1 | 7 | 1076 | |||
XM_011510750.2 | XP_011509052.1 | 5 | 855 | |||
XM_017003470.1 | XP_016858959.1 | 7 | 1073 | |||
XM_017003471.1 | XP_016858960.1 | 7 | 1076 | |||
XM_017003472.1 | XP_016858961.1 | 6 | 1009 | |||
XM_017003473.1 | XP_016858962.1 | 4 | 737 | |||
GenBank mRNA | AL833869.1 | - | 5 | 760 | ||
BC064511.1 | - | 6 | 923 | 6644987 |
Gene Symbol: FAHD2CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003698.1 | 2 | 269 | |||
GenBank mRNA | BI916267.1 | - | 2 | 236 | ||
BM790702.1 | - | 2 | 269 | |||
CN274361.1 | - | 2 | 202 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
