siRNA Details
Gene Symbol: NPIPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310136.1 | NP_001297065.1 | 4 | 562 | ||
XM_017023622.1 | XP_016879111.1 | 5 | 686 | |||
XM_017023623.1 | XP_016879112.1 | 5 | 686 | |||
XM_017023624.1 | XP_016879113.1 | 5 | 686 | |||
XM_017023625.1 | XP_016879114.1 | 5 | 666 | |||
XM_017023626.1 | XP_016879115.1 | 6 | 1305 | |||
XM_017023627.1 | XP_016879116.1 | 5 | 686 |
Gene Symbol: LOC105376752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522797.2 | XP_011521099.1 | 4 | 531 |
Gene Symbol: NPIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006985.2 | NP_008916.2 | 4 | 400 | ||
XM_011522748.2 | XP_011521050.1 | 4 | 497 | |||
XM_011522749.2 | XP_011521051.1 | 5 | 609 | |||
XM_011522750.2 | XP_011521052.1 | 4 | 523 | |||
XM_011522751.1 | XP_011521053.1 | 4 | 496 | |||
XM_017023891.1 | XP_016879380.1 | 5 | 609 | |||
GenBank mRNA | AF132984.1 | - | 4 | 400 | ||
AK160377.1 | - | 23 | 4587 | |||
KF175165.1 | - | 4 | 436 | |||
KF175166.1 | - | 4 | 436 | |||
KF175167.1 | - | 4 | 436 | |||
KF175169.1 | - | 4 | 400 | |||
KF175209.1 | - | 5 | 618 | |||
KF175225.1 | - | 4 | 400 |
Gene Symbol: NPIPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277324.1 | NP_001264253.1 | 4 | 400 | ||
XM_005255489.3 | XP_005255546.1 | 6 | 2819 | |||
XM_005255492.4 | XP_005255549.1 | 4 | 804 | |||
XM_006720917.3 | XP_006720980.2 | 6 | 2863 | |||
XM_011522587.1 | XP_011520889.1 | 6 | 2863 | |||
XM_011522593.1 | XP_011520895.1 | 6 | 2863 | |||
XM_011522594.2 | XP_011520896.1 | 6 | 2863 | |||
XM_011522595.1 | XP_011520897.1 | 6 | 2863 | |||
XM_011522596.1 | XP_011520898.1 | 4 | 427 | |||
XM_011522597.1 | XP_011520899.1 | 6 | 2863 |
Gene Symbol: NPIPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277323.1 | NP_001264252.1 | 4 | 400 | ||
XM_005255484.3 | XP_005255541.1 | 5 | 488 | |||
XM_005255485.4 | XP_005255542.1 | 4 | 804 | |||
XM_011522576.2 | XP_011520878.2 | 6 | 2853 | |||
XM_011522577.2 | XP_011520879.2 | 6 | 2774 | |||
XM_011522578.2 | XP_011520880.2 | 4 | 496 | |||
XM_011522580.2 | XP_011520882.2 | 4 | 427 | |||
XM_011522581.1 | XP_011520883.1 | 6 | 2863 | |||
XM_011522583.2 | XP_011520885.1 | 6 | 2863 | |||
XM_011522584.1 | XP_011520886.1 | 6 | 2863 | |||
XM_011522585.1 | XP_011520887.1 | 4 | 427 | |||
XM_011522586.1 | XP_011520888.1 | 6 | 2863 | |||
XM_017023553.1 | XP_016879042.1 | 6 | 2863 | |||
XM_017023554.1 | XP_016879043.1 | 5 | 2776 |
Gene Symbol: NPIPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277325.1 | NP_001264254.1 | 4 | 400 | ||
XM_011522329.2 | XP_011520631.1 | 6 | 811 | |||
XM_011522336.2 | XP_011520638.1 | 6 | 804 | |||
XM_011522337.1 | XP_011520639.1 | 4 | 427 | |||
GenBank mRNA | CB123100.1 | - | 1 | 78 | ||
EL734608.1 | - | 4 | 400 | |||
KF175168.1 | - | 4 | 397 | |||
KF175170.1 | - | 4 | 400 | |||
KF175171.1 | - | 4 | 394 | |||
KF175172.1 | - | 4 | 400 | |||
KF175173.1 | - | 2 | 175 |
Gene Symbol: NPIPA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282507.1 | NP_001269436.1 | 4 | 400 | ||
XM_005255735.4 | XP_005255792.1 | 4 | 522 | |||
XM_011522339.2 | XP_011520641.1 | 4 | 448 | |||
XM_017022833.1 | XP_016878322.1 | 5 | 473 | |||
XM_017022834.1 | XP_016878323.1 | 5 | 525 | |||
XM_017022835.1 | XP_016878324.1 | 5 | 586 | |||
XM_017022836.1 | XP_016878325.1 | 4 | 393 | |||
XM_017022837.1 | XP_016878326.1 | 5 | 473 | |||
XM_017022838.1 | XP_016878327.1 | 4 | 868 | |||
XM_017022839.1 | XP_016878328.1 | 5 | 473 | |||
XM_017022840.1 | XP_016878329.1 | 4 | 413 | |||
GenBank mRNA | BI046418.1 | - | 1 | 51 |
Gene Symbol: NPIPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282511.1 | NP_001269440.1 | 4 | 400 | ||
XM_005255738.4 | XP_005255795.1 | 4 | 522 | |||
XM_006720830.3 | XP_006720893.1 | 4 | 1420 | |||
XM_011522342.2 | XP_011520644.1 | 4 | 449 | |||
XM_017022841.1 | XP_016878330.1 | 5 | 473 | |||
XM_017022842.1 | XP_016878331.1 | 5 | 525 | |||
XM_017022843.1 | XP_016878332.1 | 5 | 586 | |||
XM_017022844.1 | XP_016878333.1 | 4 | 393 | |||
XM_017022845.1 | XP_016878334.1 | 5 | 473 | |||
XM_017022846.1 | XP_016878335.1 | 5 | 473 | |||
XM_017022847.1 | XP_016878336.1 | 4 | 413 | |||
GenBank mRNA | BQ101079.1 | - | 4 | 446 |
Gene Symbol: NPIPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310137.1 | NP_001297066.1 | 4 | 508 | ||
GenBank mRNA | KF175204.1 | - | 4 | 451 |
Gene Symbol: NPIPB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306094.1 | NP_001293023.1 | 4 | 530 | ||
XM_011523496.2 | XP_011521798.1 | 5 | 586 | |||
XM_011523498.2 | XP_011521800.1 | 5 | 620 | |||
XM_017023248.1 | XP_016878737.1 | 5 | 1023 | |||
XM_017023249.1 | XP_016878738.1 | 4 | 840 |
Gene Symbol: NPIPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | KF175178.1 | - | 4 | 409 |
Gene Symbol: NPIPB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282524.1 | NP_001269453.1 | 4 | 600 | ||
XM_005255741.3 | XP_005255798.1 | 5 | 1412 | |||
XM_005255742.4 | XP_005255799.1 | 5 | 1158 | |||
XM_011545929.2 | XP_011544231.1 | 6 | 1232 | |||
XM_011545931.2 | XP_011544233.1 | 5 | 865 | |||
XM_017023628.1 | XP_016879117.1 | 5 | 615 | |||
XM_017023629.1 | XP_016879118.1 | 5 | 1158 | |||
XM_017023630.1 | XP_016879119.1 | 6 | 885 | |||
XM_017023631.1 | XP_016879120.1 | 4 | 1229 | |||
XM_017023632.1 | XP_016879121.1 | 5 | 615 | |||
GenBank mRNA | BC144439.1 | - | 4 | 561 | ||
BC157836.1 | - | 4 | 1250 | 9057055 | ||
KF175222.1 | - | 4 | 562 |
Gene Symbol: NPIPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287250.1 | NP_001274179.1 | 4 | 600 | ||
NM_001287251.1 | NP_001274180.1 | 5 | 686 | |||
XM_003118719.7 | XP_003118767.2 | 5 | 1161 | |||
XM_011545690.2 | XP_011543992.1 | 6 | 1250 | |||
XM_011545693.2 | XP_011543995.1 | 6 | 1249 | |||
XM_017022814.1 | XP_016878303.1 | 6 | 1250 | |||
XM_017022815.1 | XP_016878304.1 | 6 | 770 | |||
XM_017022816.1 | XP_016878305.1 | 6 | 1250 | |||
XM_017022817.1 | XP_016878306.1 | 5 | 1162 | |||
XM_017022818.1 | XP_016878307.1 | 5 | 2487 | |||
XM_017022819.1 | XP_016878308.1 | 6 | 1177 | |||
XM_017022820.1 | XP_016878309.1 | 6 | 894 | |||
XM_017022821.1 | XP_016878310.1 | 5 | 1760 | |||
XM_017022822.1 | XP_016878311.1 | 6 | 2763 | |||
XM_017022823.1 | XP_016878312.1 | 4 | 2399 | |||
XM_017022824.1 | XP_016878313.1 | 6 | 1176 | |||
XM_017022825.1 | XP_016878314.1 | 6 | 894 | |||
XM_017022826.1 | XP_016878315.1 | 5 | 711 | |||
XM_017022828.1 | XP_016878317.1 | 6 | 1249 | |||
GenBank mRNA | BC171825.1 | - | 5 | 1336 |
Gene Symbol: PKD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036447.1 | 29 | 5705 | |||
GenBank mRNA | AK124516.1 | - | 24 | 3960 | ||
AK298381.1 | - | 2 | 158 | |||
BC039707.1 | - | 5 | 719 | 4479933 | ||
BC046145.1 | - | 6 | 811 | |||
KF175174.1 | - | 4 | 436 | |||
KF175210.1 | - | 5 | 679 | |||
KF175211.1 | - | 6 | 756 | |||
KF175212.1 | - | 7 | 569 | |||
KF175214.1 | - | 5 | 618 | |||
KF175219.1 | - | 4 | 586 |
Gene Symbol: PKD1P6-NPIPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_123721.1 | 23 | 4493 | |||
NR_123722.1 | 13 | 2354 | ||||
GenBank mRNA | AK131084.1 | - | 18 | 4857 | ||
AK131085.1 | - | 17 | 4763 | |||
AK302708.1 | - | 13 | 2354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
