siRNA Details
Gene Symbol: ESF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276380.1 | NP_001263309.1 | 2 | 662 | ||
NM_016649.3 | NP_057733.2 | 2 | 673 | |||
XM_017027874.1 | XP_016883363.1 | 2 | 2125 | |||
GenBank mRNA | AK000375.1 | - | 2 | 674 | ||
AK022369.1 | - | 2 | 624 | |||
AK074299.1 | - | 1 | 159 | |||
BC020952.1 | - | 1 | 292 | |||
BC030513.1 | - | 1 | 623 | |||
BC039706.1 | - | 1 | 614 | |||
BC046107.1 | - | 1 | 612 |
