siRNA Details
Gene Symbol: NPIPB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282524.1 | NP_001269453.1 | 7 | 799 | ||
XM_005255741.3 | XP_005255798.1 | 8 | 1611 | |||
XM_005255743.3 | XP_005255800.1 | 4 | 2190 | |||
XM_011545929.2 | XP_011544231.1 | 9 | 1431 | |||
XM_011545931.2 | XP_011544233.1 | 8 | 1064 | |||
XM_017023628.1 | XP_016879117.1 | 8 | 814 | |||
XM_017023629.1 | XP_016879118.1 | 8 | 1357 | |||
XM_017023630.1 | XP_016879119.1 | 9 | 1084 | |||
XM_017023631.1 | XP_016879120.1 | 7 | 1428 | |||
XM_017023632.1 | XP_016879121.1 | 8 | 814 | |||
GenBank mRNA | BC144439.1 | - | 7 | 760 | ||
BC157836.1 | - | 7 | 1449 | 9057055 | ||
KF175222.1 | - | 7 | 761 |
Gene Symbol: LOC613037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321892.1 | NP_001308821.1 | 9 | 900 | ||
XM_017023514.1 | XP_016879003.1 | 9 | 1189 | |||
XM_017023515.1 | XP_016879004.1 | 10 | 1208 | |||
XM_017023516.1 | XP_016879005.1 | 9 | 1235 | |||
XM_017023517.1 | XP_016879006.1 | 9 | 966 | |||
XM_017023518.1 | XP_016879007.1 | 9 | 1054 | |||
XM_017023519.1 | XP_016879008.1 | 9 | 1111 | |||
XM_017023520.1 | XP_016879009.1 | 4 | 2175 | |||
XM_017023521.1 | XP_016879010.1 | 5 | 430 | |||
XM_017023522.1 | XP_016879011.1 | 2 | 839 |
Gene Symbol: NPIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006985.2 | NP_008916.2 | 8 | 707 | ||
XM_011522748.2 | XP_011521050.1 | 8 | 804 | |||
XM_011522749.2 | XP_011521051.1 | 9 | 916 | |||
XM_011522750.2 | XP_011521052.1 | 7 | 722 | |||
XM_011522751.1 | XP_011521053.1 | 9 | 873 | |||
XM_017023891.1 | XP_016879380.1 | 10 | 986 | |||
GenBank mRNA | AF132984.1 | - | 8 | 707 | ||
AK160377.1 | - | 28 | 4964 | |||
KF175208.1 | - | 8 | 854 | |||
KF175209.1 | - | 10 | 995 | |||
KF175225.1 | - | 8 | 707 |
Gene Symbol: NPIPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277324.1 | NP_001264253.1 | 8 | 707 | ||
XM_005255489.3 | XP_005255546.1 | 10 | 3126 | |||
GenBank mRNA | KF175213.1 | - | 8 | 470 |
Gene Symbol: NPIPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277323.1 | NP_001264252.1 | 8 | 707 | ||
XM_005255484.3 | XP_005255541.1 | 9 | 795 | |||
XM_011522576.2 | XP_011520878.2 | 10 | 3160 | |||
XM_011522577.2 | XP_011520879.2 | 10 | 3081 | |||
XM_011522578.2 | XP_011520880.2 | 8 | 803 | |||
XM_011522580.2 | XP_011520882.2 | 8 | 734 | |||
XM_017023553.1 | XP_016879042.1 | 10 | 3170 | |||
XM_017023554.1 | XP_016879043.1 | 9 | 3083 |
Gene Symbol: NPIPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277325.1 | NP_001264254.1 | 8 | 707 | ||
XM_011522336.2 | XP_011520638.1 | 11 | 1181 | |||
GenBank mRNA | CB123100.1 | - | 5 | 385 | ||
KF175170.1 | - | 8 | 707 | |||
KF175173.1 | - | 6 | 482 |
Gene Symbol: NPIPA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282507.1 | NP_001269436.1 | 8 | 707 | ||
XM_005255735.4 | XP_005255792.1 | 8 | 829 | |||
XM_011522339.2 | XP_011520641.1 | 8 | 755 | |||
XM_017022833.1 | XP_016878322.1 | 9 | 780 | |||
XM_017022834.1 | XP_016878323.1 | 9 | 832 | |||
XM_017022835.1 | XP_016878324.1 | 9 | 893 | |||
XM_017022836.1 | XP_016878325.1 | 8 | 700 | |||
XM_017022837.1 | XP_016878326.1 | 9 | 1643 | |||
XM_017022838.1 | XP_016878327.1 | 8 | 2038 | |||
XM_017022839.1 | XP_016878328.1 | 8 | 1535 | |||
GenBank mRNA | BI046418.1 | - | 5 | 358 | ||
BI832721.1 | - | 2 | 89 |
Gene Symbol: NPIPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282511.1 | NP_001269440.1 | 8 | 707 | ||
XM_005255738.4 | XP_005255795.1 | 8 | 829 | |||
XM_006720830.3 | XP_006720893.1 | 8 | 2590 | |||
XM_011522342.2 | XP_011520644.1 | 8 | 756 | |||
XM_017022841.1 | XP_016878330.1 | 9 | 780 | |||
XM_017022842.1 | XP_016878331.1 | 9 | 832 | |||
XM_017022843.1 | XP_016878332.1 | 9 | 893 | |||
XM_017022844.1 | XP_016878333.1 | 8 | 700 | |||
XM_017022845.1 | XP_016878334.1 | 9 | 1643 | |||
XM_017022846.1 | XP_016878335.1 | 8 | 1535 |
Gene Symbol: NPIPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310137.1 | NP_001297066.1 | 7 | 707 | ||
GenBank mRNA | KF175195.1 | - | 7 | 611 | ||
KF175204.1 | - | 7 | 650 |
Gene Symbol: NPIPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130464.2 | NP_569731.2 | 8 | 783 | ||
GenBank mRNA | AB209632.1 | - | 7 | 3350 | ||
AK128772.1 | - | 3 | 1903 | |||
AK294452.1 | - | 4 | 346 | |||
AK295170.1 | - | 8 | 782 | |||
AK300277.1 | - | 100 | ||||
AK303500.1 | - | 4 | 277 | |||
AK304162.1 | - | 6 | 565 | |||
AK316459.1 | - | 4 | 212 | |||
BC036263.1 | - | 8 | 783 | 3903371 | ||
BC061522.1 | - | 1 | 91 | 6190707 | ||
BC094882.1 | - | 7 | 710 | 3093162 | ||
BM557638.1 | - | 4 | 357 | |||
BQ721066.1 | - | 1 | 90 | |||
KF175176.1 | - | 7 | 707 | |||
KF175178.1 | - | 7 | 608 | |||
KF175179.1 | - | 7 | 650 | |||
KF175180.1 | - | 7 | 650 | |||
KF175182.1 | - | 7 | 650 | |||
KF175184.1 | - | 6 | 533 | |||
KF175185.1 | - | 7 | 650 | |||
KF175186.1 | - | 6 | 524 | |||
KF175187.1 | - | 7 | 650 | |||
KF175188.1 | - | 7 | 611 | |||
KF175189.1 | - | 7 | 611 | |||
KF175190.1 | - | 7 | 650 | |||
KF175191.1 | - | 7 | 611 | |||
KF175192.1 | - | 7 | 611 | |||
KF175193.1 | - | 7 | 614 | |||
KF175194.1 | - | 7 | 614 | |||
KF175196.1 | - | 7 | 611 | |||
KF175197.1 | - | 7 | 611 | |||
KF175198.1 | - | 7 | 707 | |||
KF175199.1 | - | 7 | 707 | |||
KF175200.1 | - | 7 | 707 | |||
KF175201.1 | - | 7 | 650 | |||
KF175202.1 | - | 7 | 611 | |||
KF175203.1 | - | 7 | 707 | |||
KF175205.1 | - | 7 | 707 | |||
KF175206.1 | - | 7 | 707 | |||
KF175215.1 | - | 9 | 969 | |||
KF175216.1 | - | 8 | 984 | |||
KF175218.1 | - | 6 | 721 | |||
KF175223.1 | - | 7 | 707 | |||
KF175224.1 | - | 7 | 650 |
Gene Symbol: NPIPB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310148.1 | NP_001297077.1 | 8 | 932 | ||
XM_017023244.1 | XP_016878733.1 | 7 | 2943 | |||
XM_017023245.1 | XP_016878734.1 | 8 | 2780 | |||
XM_017023246.1 | XP_016878735.1 | 8 | 2701 | |||
XM_017023247.1 | XP_016878736.1 | 7 | 934 |
Gene Symbol: NPIPB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135865.1 | NP_001129337.1 | 7 | 986 | ||
GenBank mRNA | AK296301.1 | - | 267 | |||
AK296338.1 | - | 7 | 827 | |||
AK303264.1 | - | 4 | 299 | |||
AK303572.1 | - | 1 | 368 |
Gene Symbol: NPIPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310136.1 | NP_001297065.1 | 7 | 761 | ||
XM_017023622.1 | XP_016879111.1 | 8 | 1032 | |||
XM_017023623.1 | XP_016879112.1 | 8 | 1032 | |||
XM_017023624.1 | XP_016879113.1 | 8 | 885 | |||
XM_017023625.1 | XP_016879114.1 | 8 | 865 | |||
XM_017023626.1 | XP_016879115.1 | 9 | 1504 |
Gene Symbol: NPIPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287250.1 | NP_001274179.1 | 7 | 799 | ||
NM_001287251.1 | NP_001274180.1 | 8 | 885 | |||
XM_003118719.7 | XP_003118767.2 | 8 | 1360 | |||
XM_011545690.2 | XP_011543992.1 | 10 | 1521 | |||
XM_011545693.2 | XP_011543995.1 | 9 | 1448 | |||
XM_017022814.1 | XP_016878303.1 | 9 | 1596 | |||
XM_017022815.1 | XP_016878304.1 | 9 | 1116 | |||
XM_017022816.1 | XP_016878305.1 | 9 | 1596 | |||
XM_017022817.1 | XP_016878306.1 | 8 | 1508 | |||
XM_017022818.1 | XP_016878307.1 | 8 | 2833 | |||
XM_017022819.1 | XP_016878308.1 | 9 | 1523 | |||
XM_017022820.1 | XP_016878309.1 | 9 | 1240 | |||
XM_017022821.1 | XP_016878310.1 | 8 | 2106 | |||
XM_017022822.1 | XP_016878311.1 | 9 | 3109 | |||
XM_017022823.1 | XP_016878312.1 | 7 | 2745 | |||
XM_017022824.1 | XP_016878313.1 | 9 | 1375 | |||
XM_017022825.1 | XP_016878314.1 | 9 | 1093 | |||
XM_017022826.1 | XP_016878315.1 | 8 | 1057 | |||
XM_017022827.1 | XP_016878316.1 | 4 | 804 | |||
GenBank mRNA | BC171825.1 | - | 8 | 1535 |
Gene Symbol: PDXDC2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003610.1 | 26 | 3813 | |||
GenBank mRNA | AL833251.1 | - | 26 | 3813 | ||
BC063633.1 | - | 24 | 2300 | |||
BX647358.1 | - | 2 | 5377 | |||
KF175207.1 | - | 7 | 656 | |||
KF175220.1 | - | 8 | 631 | |||
KF175221.1 | - | 7 | 635 |
Gene Symbol: PKD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036447.1 | 31 | 5843 | |||
GenBank mRNA | AF229069.1 | - | 1 | 66 | ||
AK124516.1 | - | 26 | 4098 | |||
BC039707.1 | - | 10 | 1096 | 4479933 | ||
BC046145.1 | - | 10 | 1118 | |||
KF175210.1 | - | 10 | 1059 | |||
KF175211.1 | - | 11 | 1133 | |||
KF175212.1 | - | 11 | 876 | |||
KF175214.1 | - | 9 | 925 | |||
KF175219.1 | - | 9 | 963 |
Gene Symbol: PKD1P6-NPIPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_123721.1 | 28 | 4870 | |||
GenBank mRNA | AK131084.1 | - | 23 | 5234 | ||
AK131085.1 | - | 22 | 5140 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
