siRNA Details
Gene Symbol: CRIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005249103.3 | XP_005249160.1 | 1 | 1103 | ||
XM_011514609.2 | XP_011512911.1 | 1 | 1103 | |||
XM_017010856.1 | XP_016866345.1 | 1 | 1103 | |||
XM_017010857.1 | XP_016866346.1 | 1 | 1103 |
Gene Symbol: SLC50A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BI711397.1 | - | 1 | 275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
