siRNA Details
Gene Symbol: CYP4F35P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026756.1 | 4 | 2805 | |||
GenBank mRNA | BC037172.1 | - | 4 | 2805 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
