siRNA Details
Gene Symbol: ENAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008493.1 | NP_001008493.1 | 15 | 9552 | ||
NM_018212.4 | NP_060682.2 | 14 | 9489 | |||
XM_005273187.2 | XP_005273244.1 | 16 | 9592 | |||
XM_017001746.1 | XP_016857235.1 | 17 | 10827 | |||
XM_017001751.1 | XP_016857240.1 | 14 | 9837 | |||
XM_017001753.1 | XP_016857242.1 | 18 | 9631 | |||
XM_017001754.1 | XP_016857243.1 | 17 | 9575 | |||
XM_017001755.1 | XP_016857244.1 | 17 | 9318 | |||
XM_017001756.1 | XP_016857245.1 | 15 | 9249 | |||
XM_017001757.1 | XP_016857246.1 | 15 | 9152 |
Gene Symbol: ARID3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC060824.1 | - | 4 | 1517 |
Gene Symbol: C19orf60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054670.1 | - | 4 | 1088 | ||
AK091925.1 | - | 3 | 1391 | |||
AK094710.1 | - | 1 | 1581 |
Gene Symbol: EHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124613.1 | - | 1 | 404 |
Gene Symbol: EXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123414.1 | - | 1 | 19 |
Gene Symbol: LOC100288254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130933.1 | 2 | 1378 | |||
GenBank mRNA | AK097772.1 | - | 2 | 1302 |
Gene Symbol: PLA2R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005246392.3 | XP_005246449.1 | 31 | 6890 | ||
XM_011510820.2 | XP_011509122.1 | 31 | 6536 |
Gene Symbol: PPM1K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152542.4 | NP_689755.3 | 7 | 5054 | ||
XM_006714111.3 | XP_006714174.1 | 7 | 5115 | |||
XM_017007803.1 | XP_016863292.1 | 7 | 4702 | |||
XM_017007804.1 | XP_016863293.1 | 8 | 5242 | |||
XM_017007805.1 | XP_016863294.1 | 9 | 5242 | |||
XM_017007806.1 | XP_016863295.1 | 7 | 4575 | |||
XM_017007807.1 | XP_016863296.1 | 6 | 4293 | |||
GenBank mRNA | AK092450.1 | - | 1 | 211 |
Gene Symbol: SAMM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093144.1 | - | 1 | 1569 |
Gene Symbol: TMEM120B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123181.1 | - | 2 | 342 |
Gene Symbol: TRAPPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011658.3 | NP_001011658.1 | 6 | 1079 | ||
NM_001128835.2 | NP_001122307.2 | 6 | 1073 | |||
NM_014563.5 | NP_055378.1 | 5 | 937 | |||
XM_011545565.1 | XP_011543867.1 | 7 | 1147 | |||
XM_011545566.2 | XP_011543868.1 | 6 | 1003 | |||
GenBank mRNA | AK055671.1 | - | 1 | 244 | ||
BC016915.1 | - | 5 | 900 | 4432196 | ||
BC052618.1 | - | 5 | 900 | 6581382 |
Gene Symbol: USP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127055.1 | - | 1 | 193 |
Gene Symbol: XPNPEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022098.3 | NP_071381.1 | 10 | 5458 |
Gene Symbol: ZNF818P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073396.1 | 1 | 1274 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
