siRNA Details
Gene Symbol: FAM21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005751.2 | NP_001005751.1 | 23 | 2544 | ||
NM_001291398.1 | NP_001278327.1 | 23 | 2544 | |||
XM_005269806.3 | XP_005269863.2 | 22 | 2420 | |||
XM_005269807.3 | XP_005269864.2 | 22 | 2405 | |||
XM_005269808.3 | XP_005269865.2 | 23 | 2492 | |||
XM_005269810.3 | XP_005269867.2 | 5 | 637 | |||
XM_006717831.3 | XP_006717894.2 | 22 | 2339 | |||
XM_006717832.3 | XP_006717895.2 | 21 | 2267 | |||
XM_011539786.2 | XP_011538088.1 | 23 | 2492 | |||
XM_017016227.1 | XP_016871716.1 | 23 | 2492 | |||
XM_017016228.1 | XP_016871717.1 | 22 | 2420 | |||
XM_017016229.1 | XP_016871718.1 | 22 | 2405 | |||
XM_017016230.1 | XP_016871719.1 | 22 | 2357 | |||
XM_017016231.1 | XP_016871720.1 | 22 | 2339 | |||
XM_017016232.1 | XP_016871721.1 | 21 | 2333 | |||
XM_017016233.1 | XP_016871722.1 | 20 | 2067 | |||
XM_017016234.1 | XP_016871723.1 | 22 | 2427 | |||
XM_017016235.1 | XP_016871724.1 | 14 | 1443 | |||
GenBank mRNA | AK001686.1 | - | 20 | 2196 | ||
AK304116.1 | - | 23 | 2480 | |||
BC075815.1 | - | 20 | 2109 | |||
BC082258.1 | - | 23 | 2471 | 6278433 | ||
BC144479.1 | - | 23 | 2573 | 9053006 | ||
CX762870.1 | - | 2 | 300 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
