siRNA Details
Gene Symbol: KCNQ1OT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002728.3 | 1 | 63540 |
Gene Symbol: LINC00504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126435.1 | 7 | 1674 | |||
GenBank mRNA | BC036758.2 | - | 7 | 1686 | 5563071 |
Gene Symbol: ROR1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110665.1 | 4 | 692 | |||
GenBank mRNA | AK096291.1 | - | 5 | 781 |
Gene Symbol: SMC5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR936772.1 | - | 3 | 5336 |
Gene Symbol: UGDH-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047679.1 | 5 | 1552 | |||
GenBank mRNA | AK056558.1 | - | 5 | 1372 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
