siRNA Details
Gene Symbol: ATP9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK295981.1 | - | 5 | 615 | ||
BC053561.1 | - | 5 | 533 |
Gene Symbol: EYS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647741.1 | - | 5 | 4923 |
Gene Symbol: KCNQ1OT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002728.3 | 1 | 63929 |
Gene Symbol: LINC00504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126435.1 | 7 | 2062 | |||
GenBank mRNA | BC036758.2 | - | 7 | 2074 | 5563071 |
Gene Symbol: SMC5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR936772.1 | - | 3 | 5719 |
Gene Symbol: TBC1D8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648230.1 | - | 7 | 3085 |
Gene Symbol: UGDH-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047679.1 | 5 | 1939 |

More Information
Other Species Targeted

