siRNA Details
Gene Symbol: XAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017523.3 | NP_059993.2 | 7 | 2407 | ||
NM_199139.2 | NP_954590.1 | 6 | 2350 | |||
NR_046396.1 | 6 | 2321 | ||||
NR_046397.1 | 5 | 2264 | ||||
NR_046398.1 | 5 | 2154 | ||||
XM_005256706.4 | XP_005256763.1 | 7 | 2979 | |||
XM_005256708.2 | XP_005256765.1 | 8 | 2969 | |||
XM_011523945.2 | XP_011522247.1 | 8 | 2794 | |||
XM_011523947.2 | XP_011522249.1 | 7 | 2315 | |||
XM_017024780.1 | XP_016880269.1 | 7 | 2608 | |||
GenBank mRNA | AK091799.1 | - | 1 | 761 | ||
BX648588.1 | - | 6 | 2737 | |||
BX649188.1 | - | 7 | 4654 |
Gene Symbol: BRDTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124282.1 | - | 1 | 1680 |
Gene Symbol: CFAP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC041426.1 | - | 7 | 1590 |
Gene Symbol: CYSLTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK023929.1 | - | 2 | 1667 |
Gene Symbol: DNAH12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011533471.2 | XP_011531773.1 | 59 | 9672 |
Gene Symbol: ESR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M69297.1 | - | 3 | 1140 |
Gene Symbol: FSIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092099.1 | - | 8 | 2354 |
Gene Symbol: GMDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011514502.2 | XP_011512804.1 | 10 | 1358 |
Gene Symbol: HELLPAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | JX088243.1 | - | 1 | 42677 |
Gene Symbol: LCLAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095284.1 | - | 7 | 1530 |
Gene Symbol: LINC00504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126435.1 | 7 | 2795 | |||
GenBank mRNA | BC036758.2 | - | 7 | 2807 | 5563071 |
Gene Symbol: LINC00976 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BF691581.1 | - | 312 |
Gene Symbol: LOC100129434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125368.1 | 3 | 2572 | |||
GenBank mRNA | AK096196.1 | - | 3 | 2781 |
Gene Symbol: LOC101926898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125837.1 | 3 | 1048 | |||
GenBank mRNA | BC042616.1 | - | 3 | 1050 |
Gene Symbol: LOC101928137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110130.1 | 4 | 1235 | |||
GenBank mRNA | AK310448.1 | - | 4 | 1237 |
Gene Symbol: LOC101928526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC033553.1 | - | 1 | 2278 |
Gene Symbol: LOC101929163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123889.1 | - | 2 | 3377 |
Gene Symbol: LOC105371828 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126827.1 | - | 2 | 1355 |
Gene Symbol: NLRP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK310900.1 | - | 12 | 3696 |
Gene Symbol: ODF2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007022.2 | NP_001007023.2 | 18 | 3219 | ||
NM_020729.2 | NP_065780.2 | 18 | 3178 | |||
XM_011541819.2 | XP_011540121.1 | 18 | 3224 | |||
XM_011541821.2 | XP_011540123.1 | 17 | 3065 | |||
XM_017001879.1 | XP_016857368.1 | 17 | 3137 | |||
GenBank mRNA | AB033055.1 | - | 18 | 3139 | ||
BC047368.1 | - | 18 | 3117 | 5276959 |
Gene Symbol: TBC1D8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648230.1 | - | 7 | 3819 |
Gene Symbol: TCF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647333.1 | - | 1 | 6572 |
Gene Symbol: TMEM212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164436.1 | NP_001157908.1 | 5 | 995 | ||
GenBank mRNA | AK026825.1 | - | 5 | 975 |
Gene Symbol: UGDH-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047679.1 | 5 | 2673 | |||
GenBank mRNA | AK056558.1 | - | 5 | 2493 |
Gene Symbol: WDPCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX538331.1 | - | 14 | 2307 |
Gene Symbol: XRCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005431.1 | NP_005422.1 | 3 | 2022 | ||
GenBank mRNA | AF035587.1 | - | 3 | 2022 | ||
CR749256.1 | - | 3 | 1956 |
Gene Symbol: ZNF480 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297624.1 | NP_001284553.1 | 4 | 3487 | ||
NM_001297625.1 | NP_001284554.1 | 4 | 3489 | |||
NM_144684.3 | NP_653285.2 | 5 | 3616 | |||
XM_011526465.2 | XP_011524767.1 | 5 | 4018 | |||
GenBank mRNA | AL832560.1 | - | 4 | 3466 | ||
AY512662.1 | - | 5 | 3585 | |||
BC018783.1 | - | 5 | 3585 | 4873910 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
