siRNA Details
Gene Symbol: DBET | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121644.1 | 1 | 2368 | |||
GenBank mRNA | JQ639078.1 | - | 3 | 2364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
