siRNA Details
Gene Symbol: DDX19A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209401.1 | - | 1 | 259 |
Gene Symbol: ANGPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK225698.1 | - | 8 | 2321 |
Gene Symbol: ANKLE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005266160.1 | XP_005266217.1 | 1 | 37 | ||
XM_017019076.1 | XP_016874565.1 | 1 | 37 |
Gene Symbol: CAPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090469.1 | - | 1 | 1164 |
Gene Symbol: INTS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017018560.1 | XP_016874049.1 | 23 | 4969 |
Gene Symbol: NOA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128696.1 | - | 1 | 524 |
Gene Symbol: PPP3CB-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132103.1 | 3 | 442 | |||
NR_132104.1 | 3 | 442 | ||||
GenBank mRNA | BU150385.1 | - | 2 | 236 | ||
DA883449.1 | - | 2 | 390 |
Gene Symbol: PSORS1C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB933320.1 | - | 3 | 564 | ||
LC050981.1 | - | 3 | 590 | |||
LC050986.1 | - | 3 | 590 | |||
LC050987.1 | - | 4 | 857 |
Gene Symbol: SALL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318031.1 | NP_001304960.1 | 4 | 2411 | ||
NM_020436.4 | NP_065169.1 | 4 | 3722 | |||
GenBank mRNA | CN271900.1 | - | 1 | 143 |
Gene Symbol: SLC11A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000578.3 | NP_000569.3 | 15 | 2629 | ||
XM_005246793.3 | XP_005246850.1 | 14 | 2749 | |||
XM_005246794.3 | XP_005246851.1 | 16 | 2966 | |||
XM_006712709.3 | XP_006712772.1 | 15 | 2823 | |||
XM_006712710.3 | XP_006712773.1 | 14 | 2700 | |||
XM_006712711.3 | XP_006712774.1 | 13 | 2626 | |||
XM_011511684.2 | XP_011509986.1 | 16 | 3001 | |||
XM_011511685.2 | XP_011509987.1 | 15 | 2858 | |||
XM_017004765.1 | XP_016860254.1 | 14 | 2769 | |||
XM_017004766.1 | XP_016860255.1 | 15 | 2860 | |||
XM_017004767.1 | XP_016860256.1 | 12 | 2087 | |||
GenBank mRNA | AB209650.1 | - | 13 | 5209 | ||
BC033754.1 | - | 14 | 2063 | |||
D38171.1 | - | 13 | 2140 | |||
D50402.1 | - | 15 | 2468 | |||
D50403.1 | - | 15 | 2379 |
Gene Symbol: TBC1D5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014744.2 | NP_055559.1 | 1 | 1104 | ||
GenBank mRNA | D86965.1 | - | 1 | 1103 |
Gene Symbol: TXNDC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX538131.1 | - | 5 | 2386 |
Gene Symbol: YIPF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095555.1 | - | 3 | 1555 |
Gene Symbol: ZNF880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017026812.1 | XP_016882301.1 | 5 | 2443 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
