siRNA Details
Gene Symbol: RPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002952.3 | NP_002943.2 | 1 | 913 | ||
GenBank mRNA | AB065088.1 | - | 1 | 557 | ||
AB065089.1 | - | 5 | 1157 | |||
AB082925.1 | - | 1 | 766 | |||
BC001795.1 | - | 1 | 901 | 3353508 | ||
BC004520.2 | - | 1 | 302 | |||
BC006559.2 | - | 1 | 901 | 2959602 | ||
BC008329.2 | - | 1 | 894 | |||
BC008862.2 | - | 1 | 901 | 4126765 | ||
BC010165.2 | - | 1 | 896 | 4040679 | ||
BC012354.1 | - | 1 | 896 | 4564801 | ||
BC013833.1 | - | 1 | 895 | |||
BC016178.1 | - | 6 | 893 | 3909865 | ||
BC016951.2 | - | 1 | 901 | 3883854 | ||
BC018993.2 | - | 1 | 896 | 4127411 | ||
BC019021.2 | - | 1 | 886 | |||
BC021545.1 | - | 1 | 901 | 3538227 | ||
BC023541.1 | - | 1 | 896 | 4120795 | ||
BC025677.1 | - | 1 | 894 | 5203563 | ||
BC029979.1 | - | 1 | 901 | |||
BC032129.2 | - | 1 | 901 | 5019977 | ||
BC035427.1 | - | 1 | 896 | |||
BC066321.1 | - | 1 | 913 | 5296344 | ||
BC068051.1 | - | 1 | 902 | 4853351 | ||
BC071673.1 | - | 1 | 901 | 4540360 | ||
BC071922.1 | - | 1 | 895 | 5455280 | ||
BC071923.1 | - | 1 | 900 | 5477295 | ||
BC071924.1 | - | 1 | 896 | 6729920 | ||
BC073966.1 | - | 1 | 900 | 6047887 | ||
BC075830.1 | - | 1 | 901 | 5441508 | ||
BC103756.1 | - | 1 | 926 | 6008803 | ||
BC105985.1 | - | 1 | 902 | 5114688 | ||
BC106060.1 | - | 1 | 901 | 6471612 | ||
X17206.1 | - | 7 | 914 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
