siRNA Details
Gene Symbol: OR7E37P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002163.3 | 1 | 492 |
Gene Symbol: DAPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HQ179936.1 | - | 4 | 490 | ||
HQ179937.1 | - | 5 | 582 |
Gene Symbol: MGC72080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002822.3 | 4 | 603 | |||
GenBank mRNA | BC029615.1 | - | 4 | 613 | ||
BC042035.1 | - | 3 | 542 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
