siRNA Details
Gene Symbol: COL4A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287758.1 | NP_001274687.1 | 2, 3 | 300 | ||
NM_001287759.1 | NP_001274688.1 | 2, 3 | 300 | |||
NM_001287760.1 | NP_001274689.1 | 2, 3 | 300 | |||
NM_001847.3 | NP_001838.2 | 2, 3 | 195 | |||
NM_033641.3 | NP_378667.1 | 2, 3 | 300 | |||
XM_006724617.3 | XP_006724680.1 | 2, 3 | 189 | |||
XM_011530852.2 | XP_011529154.1 | 2, 3 | 189 | |||
XM_011530854.2 | XP_011529156.1 | 2, 3 | 190 | |||
GenBank mRNA | BC000874.1 | - | 2, 3 | 234 | ||
BC005305.1 | - | 2, 3 | 195 | |||
BC070203.1 | - | 2, 3 | 285 | |||
BC140855.1 | - | 2, 3 | 196 | 9021723 | ||
BC144669.1 | - | 2, 3 | 196 | 9053200 | ||
BT007228.1 | - | 1, 2 | 69 | |||
CN313347.1 | - | 2, 3 | 303 | |||
D21337.1 | - | 2, 3 | 300 | |||
L22763.1 | - | 2, 3 | 178 | |||
U04845.1 | - | 2, 3 | 178 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
